You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Shakers
▸
Shaker Accessories
▸
Accessories for Digital Rockers and Waving Rotators
Thermo Scientific™
Accessories for Digital Rockers and Waving Rotators
Get assorted rubberized accessories for use with Digital Rockers and Digital Waving Rotators
Have Questions?
Change view
Catalog Number
Description
88882103
Rubber Strips, 6 Each
88882101
Rubber Mat
88882102
Dimpled Rubber Mat
88882128
Rubber tube replacement
4 Options
Catalog number
88882103
Price (USD)
10.12
Online exclusive
11.90
Save 1.78
Pack of 1
Add to cart
Description:
Rubber Strips, 6 Each
Price (USD)
10.12
Online exclusive
11.90
Save 1.78
Pack of 1
Add to cart
Assorted rubberized accessories for use with Digital Rockers and Digital Waving Rotators
Specifications
Description
Rubber Strips, 6 Each
Dimensions (L x W)
180 x 7 mm
For Use With (Equipment)
Digital Rockers, Digital Waving Rotator
Material
Rubber
Packaging Quantity
6/Pk.
Product Type
Rubber Strips
Unit Size
Pack of 1