1-Step Human High-Yield Maxi IVT Kit, 2 x 2 mL - FAQs
1-Step Human High-Yield Maxi IVT Kit, 2 x 2 mL - FAQs
View additional product information for 1-Step Human High-Yield IVT Kit - FAQs (88890, 88892, 88891)
3 product FAQs found
When comparing 1-Step Human High-Yield Mini IVT Kit with 1-Step Human Coupled IVT Kit - DNA, is one faster than the other?
No, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA can be run overnight (depends upon the gene).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Do both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein?
Yes, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I'm using the 1-Step Human High-Yield Mini IVT Kit. Can I get the vector map for the pT7CFE1-NHis-GST-CHA Vector from the kit?
The pT7CFE1-NHis-GST-CHA vector is also offered as a standalone product (Cat. No. 88871) and the vector information can be accessed on the product page (https://www.thermofisher.com/order/catalog/product/88871), under "Vector information" at the bottom of the page.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.