IntelliStack™ Incubator Shaker Accessories
Thermo Scientific™
IntelliStack™ Incubator Shaker Accessories
The Thermo Scientific™ IntelliStack™ accessories are designed to work with the IntelliStack Incubator Shaker and IntelliStack CO2 Incubator Shaker.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
88900003StandIntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers w/ Drawers and Closed Sides
88900000StandIntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers
88900001Humidity and Drain Carboy KitIntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers
88900002Window CoverIntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers
Catalog number 88900003
Price (USD)
-
Product Type:
Stand
For Use With (Equipment):
IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers w/ Drawers and Closed Sides
The Thermo Scientific™ IntelliStack™ accessories are designed to work with the IntelliStack Incubator Shaker and IntelliStack CO2 Incubator Shaker.
  • IntelliStack™ Incubator Accessories are compatible with both the CO2 Incubator Shaker and the Non-CO2 Incubator Shaker.
  • The IntelliStack Stands are available with either an open support design (88900000) or with closed sides and drop down access doors (88900003). Both stand options are 34 cm in height and can be used with one or two Shakers.
  • The IntelliStack Window Cover is a removable metal visor that attaches to the shaker to protect the interior for ambient light (88900002).
  • The IntelliStack Humidity and Drain Carboy Kit maximize the humidity control capabilities of the IntelliStack Shaker.
Specifications
Dimensions (L x W x H)34.9 x 55.1 x 26.8 in. (886 x 1400 x 680 mm)
For Use With (Equipment)IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers w/ Drawers and Closed Sides
HoldsSupports up to 2 IntelliStack Shakers
Product TypeStand
Unit SizeEach