alamarBlue™ Cell Viability Assay Reagent - FAQs
alamarBlue™ Cell Viability Assay Reagent - FAQs
View additional product information for alamarBlue™ Cell Viability Assay Reagent - FAQs (88952)
2 product FAQs found
Can other cell viability reagents be used with AlgiMatrix Matrix?
Other than the Invitrogen alamarBlue Cell Viability Reagent, AlgiMatrix Matrix is also compatible with Invitrogen PrestoBlue Cell Viability Reagent and Invitrogen CyQuant Direct Cell Proliferation Assay. The protocol is almost the same as the 2D cell culture, but it needs the blank AlgiMatrix Matrix as a blank control.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the function of the Invitrogen alamarBlue reagent assay?
The Invitrogen alamarBlue reagent assay is a useful biological assay that monitors the health and function of cells by monitoring their reducing functions.