How should I prepare and store Lysozyme (Cat. No. 89833)?
Lysozyme powder can be dissolved in 0.1 M KH2PO4 (pH 6.24) to the desired concentration - up to 5 mg/mL. Once the solution has been prepared, it can be stored at 4 degrees C for up to 5-6 weeks, or at room temperature for up to 6-7 days, without affecting performance.
Find additional tips, troubleshooting help, and resources within our Cell Lysis and Fractionation Support Center.
Find additional tips, troubleshooting help, and resources within our Cell Lysis and Fractionation Support Center.