This product is only available through your local Thermo Scientific supplier. Learn where to buy >

Zeba™ Spin Desalting Columns, 7K MWCO, 0.5 mL

Catalog number: 89883B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Zeba Spin Desalting Columns, 7K MWCO, 0.5 mL are polypropylene devices containing a proprietary high-performance size-exclusion chromatography resin that provides an excellent protein desalting and recovery in a centrifuge format.

Features of Zeba Spin Desalting Columns, 7K MWCO, 0.5 mL:

High recovery—low-binding resin maximizes protein recovery
Fast—no fraction screening or waiting for protein to emerge by gravity-flow
Easy-to-use—no cumbersome column preparation or equilibration
Flexible—available in spin columns, filter spin plates and cartridges for a range of needs
Economical—great performance at lower cost than other commercially available cartridges

Even very dilute (25 µg/mL) protein samples can be successfully processed to obtain greater than 95% retention (removal) of salts and other small molecules (< 1000 MW) and good recovery of proteins and other macromolecules (> 7000 MW).

Available Formats:
Spin Columns—75 µL, 0.5 mL (microcentrifuge), 2 mL, 5 mL, 10 mL columns
Chromatography Cartridges—compatible with automated liquid chromatography systems and with syringe processing (learn more about our Chromatography Cartridges)
96-well Spin Plates—pre-dispensed 96-well filter plates, compatible with centrifugation for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations.

More Product Data
Benchmarking experiments with Zeba Spin Desalting Columns
Chromatography cartridges for desalting and affinity purification

Related Products
Zeba™ Micro Spin Desalting Columns, 7K MWCO, 75 µL
Zeba™ Spin Desalting Columns, 7K MWCO, 2 mL
Zeba™ Spin Desalting Columns, 7K MWCO, 5 mL
Zeba™ Spin Desalting Columns, 7K MWCO, 10 mL
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Proprietary Resin, Size-Exclusion
Cut-Off or Exclusion Limit: 7.0 kDa
Format: Microcentrifuge, Spin Column
Product Line: Zeba™
Product Size: Custom

Contents & storage

Upon receipt store at 4°C.

Documents

Manuals & protocols