Sodium Deoxycholate Detergent
Thermo Scientific™

Sodium Deoxycholate Detergent

Water-soluble, ionic detergent used in cell lysis buffers (e.g., RIPA buffer) to disrupt protein interactions for liposome and membrane protein isolation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8990525 g
899045 g
Catalog number 89905
Price (USD)
526.65
Online exclusive
620.00
Save 93.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
526.65
Online exclusive
620.00
Save 93.35
Each

Thermo Scientific™ Sodium Deoxycholate is an ionic detergent that is especially useful for disrupting and dissociating protein interactions.

Features of Sodium Deoxycholate:

  • Popular anionic, bile-acid detergent for many laboratory uses
  • Effective in disrupting and dissociating many types of protein interaction
  • Can be removed from solution by dialysis
  • Useful for elution or regeneration of certain kinds of affinity columns
  • High-purity compound with low UV absorptivity

Properties of Sodium Deoxycholate:

  • Alternative Names: Sodium deoxycholic acid; deoxycholate, sodium salt
  • Chemical Name: 3, 12-α-Dihydroxy-5β-cholan-24-oic acid, monosodium salt
  • Molecular Weight: 414.6 g
  • Detergent Class: Ionic (anionic)
  • Aggregation Number: 5 (average)
  • Micelle Molecular Weight: 2000 g (average)
  • Critical Micelle Concentration (CMC): 2 to 6 mM (0.083 to 0.249%, w/v)
  • Cloud Point: Unknown
  • Dialyzable: Yes

Specifications for Sodium Deoxycholate (Part No. 89904, 89905):

  • Formula: C24H39O4Na
  • Molecular Weight: 414.6 g
  • Purity (by HPLC): ≥98%
  • Absorbance (1% Detergent Solution): 340 nm <0.02; 280 nm <0.04; 260 nm <0.06
  • pH (1% Solution): 5 to 9
  • Solubility (in water at 20°C): ≥5%

Sodium deoxycholate (deoxycholic acid) is a water-soluble, bile-acid, ionic detergent commonly used in protein methods. It is most frequently used as a component of cell lysis buffers (e.g., RIPA buffer), but also has been used for liposome preparation, isolation of membrane proteins and lipids, preventing non-specific binding in affinity chromatography and a cell culture media supplement.

The effectiveness of a detergent in any application depend upon the detergent's concentration. Too much or too little detergent can often have a deleterious effect. It is recommended that you examine a variety of detergent concentrations in your application. At concentrations above 2 mM, cholate will form micelles having MW ∼2000. The small micelle size allows easy removal by dialysis or gel filtration when needed. Note: removal of a detergent from a protein solution may result in protein precipitation and/or aggregation.

Sodium deoxycholate is the detergent recommended for stripping endotoxin (Lipopolysaccharide or LPS) from immobilized Polymyxin B columns. This is the recommended product for use with the Thermo Scientific Detoxi-Gel Endotoxin Removing Gel.

Related Products

Surfact-Amps™ Detergent Sampler (Cat. No. 28340)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity25 g
Reagent TypeDetergent (Pure)
FormPowder
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store at -20°C

Frequently asked questions (FAQs)

Are there mass spectrometry reagents for membrane proteins?

CHAPS (Cat. Nos. 28299, 28300), OTG (Cat. No. 28351) and Sodium Deoxycholate (Cat. Nos. 89904, 89905) are detergents often used to solubilize and extract membrane proteins from samples

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

What can the sodium deoxycholate detergent be used for?

This detergent is recommended for stripping endotoxin (lps) from immobilized Polymyxin B columns. One could use this product in conjunction with the Thermo Scientific Detoxi-Gel Endotoxin Removing Gel.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.