Probe Array Cartridge Carriers (Red) - FAQs
Probe Array Cartridge Carriers (Red) - FAQs
View additional product information for Probe Array Cartridge Carriers (Red) - FAQs (90-0356)
1 product FAQs found
Which probe array cartridge carriers are designed specifically for use with the GeneChip Hybridization Oven 645 (Cat. No. 00-0331)?
The following probe array cartridge carriers are designed for use with the GeneChip Hybridization Oven 645:
- Probe Array Cartridge Carriers (Red) (Cat. No. 90-0356)
- Probe Array Cartridge Carriers (Green) (Cat. No. 90-0357)
- Probe Array Cartridge Carriers (Purple) (Cat. No. 90-0358)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.