Connexin Antibody Sample Pack
Invitrogen™

Connexin Antibody Sample Pack

Mouse Polyclonal, Monoclonal Antibody
Have Questions?
Catalog NumberQuantity
900500
also known as 90-0500
90 μg
Catalog number 900500
also known as 90-0500
Price (USD)
400.65
Online exclusive
534.00
Save 133.35
Each
Quantity:
90 μg
Price (USD)
400.65
Online exclusive
534.00
Save 133.35
Each
Specificity: Connexin 26, 32, 43

Reactivity: Human, Mouse, Rat

Applications: See product data sheet for recommended Usage for each antibody.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AntigenConnexin
CloneSampler Pack
Gene ID (Entrez)2697
Primary or SecondaryPrimary
Product LineGeneChip
Purification MethodAffinity Purified
ReactivityHuman, Mouse, Rat
Reactivity abrvRt, Hu, Ms
Shipping ConditionWet Ice
Target GeneConnexin
Validated Application abrvWB, IHC (Frozen), ELISA
ApplicationsELISA, Immunohistochemistry (Frozen), Western Blot
ClassificationMonoclonal/Polyclonal
ConjugateUnconjugated
Host SpeciesMouse
Quantity90 μg
Regulatory StatusRUO
Target SpeciesHuman, Mouse, Rat
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).