How should I prepare my hybridization cocktail for the GeneChip S. aureus array?
If using the IVT Pico Assay to generate labeled material, please contact the technical support team at techsupport@thermofisher.com for additional information on hybridization specifics.