Applied Biosystems™

GeneChip™ Drosophila Genome 2.0 Array

Catalog number: 900533
Applied Biosystems™

GeneChip™ Drosophila Genome 2.0 Array

Catalog number: 900533
Catalog Number
900533
Unit Size
30 arrays
Price (USD)
Price: 19,080.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
2 arrays
Price (USD)
Price: 1,274.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
6 arrays
Price (USD)
Price: 3,885.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
90053330 arrays
Price: 19,080.00
Your Price:
-
9005312 arrays
Price: 1,274.00
Your Price:
-
9005326 arrays
Price: 3,885.00
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneChip™ Drosophila Genome 2.0 Array is a microarray tool for studying expression of Drosophila melanogaster transcripts.

• Provides comprehensive coverage of the transcribed Drosophila genome on a single array
• Comprised of 18,880 probe sets, analyzing over 18,500 transcripts
• Sophisticated bioinformatics tools available through the NetAffx™ Analysis Center to rapidly obtain biologically meaningful results

Array Profile
Sequences used in the design of the GeneChip Drosophila Genome 2.0 Array were selected from Flybase version 3.1. Oligonucleotide probes complementary to each corresponding sequence are synthesized in situ on the arrays. Fourteen pairs of oligonucleotide probes are used to measure the level of transcription of each sequence.

Instrument Software Requirements
• GeneChip Scanner 3000, enabled for High-Resolution Scanning*
• GeneChip Operating Software (GCOS) v1.1.1, which contains the High-Resolution Scanning Update
*GeneChip Scanner 3000 High-Resolution Update is standard on all instruments shipped starting in September 2003 with serial number series 502. Previous versions, such as serial number series 501, will require the GeneChip Scanner 3000 High-Resolution Update, Catalog Number 00-0110, to be installed.

For more information, please review the data sheet (pdf, 700 KB).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Drosophila Genome 2.0 Array
Array
Transcriptome Profiling
Number of Arrays
30 arrays
Format
Array Cartridge
Species
Drosophila
Product Line
GeneChip™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text