Can I use the GeneChip Wash Buffer A in a Fluidics station even if I can see crystals/precipitate present in the buffer?
GeneChip Wash Buffer A can be kept at room temperature (overnight) the day before the experiment. If you see crystals, we recommend equilibrating the buffer up to 4 hrs (if no external heating is used) until it reaches room temperature. You can also vortex it with a stir bar at room temperature.