B-PER™ Direct Bacterial Protein Extraction Kit

Catalog number: 90080

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific B-PER Direct Bacterial Protein Extraction Kit is an easy-to-use cell lysis reagent is a nonionic detergent-based solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation. All B-PER Reagents are compatible with downstream applications, such as affinity chromatography (e.g., immobilized metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification (e.g., Pierce BCA Protein Assay, Pierce 660nm Protein Assay). Depending on the particular application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.

Features of the B-PER Bacterial Protein Extraction Reagent:

• Tris buffered formulation with lysozyme and DNase enzymes
• Lysis of bacteria directly in cell culture media

All Thermo Scientific B-PER Protein Extraction Reagents are:
Ready to use—one-step cell lysis of gram-positive and gram-negative bacteria using a mild, nonionic detergent (proprietary) in Tris or phosphate buffer formulations
Fast and simple—just add B-PER Reagent to a bacterial pellet, incubate with mixing for 10 to 15 minutes and recover soluble proteins after pelleting the cell debris
Convenient B-PER Complete Reagent contains lysozyme and a universal nuclease in a single formulation with 4°C storage
Excellent yields—recover recombinant proteins from bacterial lysates or purify inclusion bodies to near-homogeneous levels
Flexible—B-PER Reagents are suitable for any scale of protein extraction and are available in phosphate and 1X and 2X Tris formulations, with and without enzymes
Compatible—completely compatible with addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (e.g., GST, 6xHis) and other applications

Convenient, Ready-to-use Formats
B-PER Bacterial Extraction Reagents are more effective than traditional sonication and typical homemade lysis buffers, many of which include detergents and components that interfere with downstream applications. B-PER Reagents are formulated in Tris or phosphate buffer at physiological pH. They extract native and soluble recombinant proteins and yield lysates that are directly compatible with most downstream workflows such as electrophoresis, affinity purification, immunoprecipitation, protein interaction analysis, crosslinking and protein labeling.

More Product Data
Improved, all-in-one B-PER Reagent for bacterial protein extraction

Related Products
B-PER™ Bacterial Protein Extraction Reagent
B-PER™ Complete Bacterial Protein Extraction Reagent
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Cell Lysis Buffer, Detergent Solution
Form: Liquid
Product Line: B-PER™
Product Size: 50 mL

Contents & storage

Sufficient For: 5 x 96-well plates of cultured cells
• B-PER Direct Reagent, 50 mL
• Lysozyme, 50 mg/mL, 0.5 mL
• DNase I, 2500 U/mL, 0.5 mL

Store B-PER Reagent at ambient temperature. Store Lysozyme and DNaseI at -20°C.

Documents

Manuals & protocols

Recommended products