HisPur™ Ni-NTA Chromatography Cartridges, 5 mL, 2 x 5 mL - FAQs
HisPur™ Ni-NTA Chromatography Cartridges, 5 mL, 2 x 5 mL - FAQs
View additional product information for HisPur™ Ni-NTA Chromatography Cartridges, 1 mL - FAQs (90099, 90098)
4 product FAQs found
With the rapid equilibrium dialysis (RED) system, if my compound has low solubility in aqueous buffer, can I use DMSO and if so, at what concentration?
A final concentration of 1% DMSO is acceptable and will not affect the study. While DMF has not been tested, it is expected that DMF should be similar to DMSO in terms of use.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can ProBond or Ni-NTA beads be used for large-scale preparations?
ProBond and Ni-NTA beads can be used in FPLC columns. However, the beads can only withstand low pressure (~43.5 psi max).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What should the typical protein recovery be when using the Probond Purification System or Ni-NTA Purification system?
Both systems are qualified by purifying 2 mg of myoglobin protein on a column and performing a Bradford assay. Protein recovery must be 75% or higher.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between HisPur Nickel Resin and HisPur Cobalt Resin?
Nickel has a higher affinity for histidines than does cobalt. It binds multiple histidines more tightly than cobalt, and requires more stringent conditions than is necessary for cobalt. The lower affinity of cobalt for multiple histidines typically results in less nonspecific binding of histidine-rich proteins that lack a his-tag compared to nickel resins.
HisPur Ni-NTA resin has a high capacity of up to 60 mg of 6xHis-tagged protein per milliliter. It is a versatile resin that can work under both native and denaturing conditions, and can also be used with a variety of lysis reagents and buffer additives.
HisPur Cobalt resin utilizes proprietary tetradentate chelating resin charged with cobalt. This system recovers highly purified protein with lower imidazole concentrations and has low metal leeching properties.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.