Sterile Automation Tips for Tecan™ Liquid Handling Platforms, 10µL, black, liquid sensing, filtered - FAQs
Sterile Automation Tips for Tecan™ Liquid Handling Platforms, 10µL, black, liquid sensing, filtered - FAQs
View additional product information for Sterile Automation Tips for Tecan™ Liquid Handling Platforms, 10µL, black, liquid sensing, filtered - FAQs (901011)
1 product FAQs found