Product Image
Thermo Scientific™
Reference Genomic DNA 103
Reference Genomic DNA 103 is used as a positive control in assays.
Have Questions?
Catalog NumberQuantity
9010122 reactions
Catalog number 901012
Price (USD)
121.00
Each
Quantity:
2 reactions
Price (USD)
121.00
Each
Reference Genomic DNA 103 is used as a positive control in assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2 reactions
Unit SizeEach
Contents & Storage
Store at –25°C to –15°C
Frequently asked questions (FAQs)
Does Ref103 come in the CytoScan XON Assay Kit (Cat. No. 931311)?
Reference Genomic DNA 103 is not included in the kit, but is available for purchase separately (Cat. No. 901012).
Note: A positive control is recommended for the CytoScan XON assay.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.