901013 - FAQs
901013 - FAQs
View additional product information for 901013 - FAQs (901013)
2 product FAQs found
What components are needed to run the Affymetrix Mouse Diversity Genotyping Array?
The components include: Affymetrix Mouse Diversity Genotyping Array
Affymetrix Genome-Wide Human SNP Nsp/Sty Assay kit 5.0/6.0
Affymetrix Power Tools (sed to run a modified BRLMM-P algorithm for genotype calling, GeneChip Fluidics Station 450
GeneChip Hybridization Oven 645
and GeneChip Scanner 3000 7G.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.