GeneChip™ HT MG-430 PM Array Plate - FAQs
GeneChip™ HT MG-430 PM Array Plate - FAQs
View additional product information for GeneChip™ HT MG-430 PM Array Plate - FAQs (901258)
1 product FAQs found
Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?
The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.