GeneChip™ HT HG-U133+PM Array Plate
Applied Biosystems™
GeneChip™ HT HG-U133+PM Array Plate
Description: Proven performance from the industry standard The GeneChip™ HT HG-U133+ PM 16-Array Plate enables high-throughput expression profiling of multiple자세히 알아보기
Have Questions?
카탈로그 번호수량
9014331 kit
카탈로그 번호 901433
제품 가격(KRW)
6,906,000
Each
수량:
1 kit
제품 가격(KRW)
6,906,000
Each
Description: Proven performance from the industry standard
The GeneChip™ HT HG-U133+ PM 16-Array Plate enables high-throughput expression profiling of multiple samples at a time using the same content as the industry-standard GeneChip Human Genome U133 Plus 2.0 Array*.
The GeneChip HT HG-U133+ PM Array Plate enables you to:
• Measure gene expression of more than 47,000 transcripts and variants, including more than 33,000 well-characterized genes and UniGene clusters per sample
• Get accurate and reproducible data by using multiple independent measurements for each transcript
Scatter plots represent median MAQC B - MAQC A signals for each array type. The high correlation coefficients indicate similar biology is being observed (Fig. 1 and Fig. 2).
Key Benefits:
• Enables increased productivity and efficiency through parallel processing
   - Process 16, 24, or 96 samples on a single array plate
• Same industry-leading performance and content as the Human Genome U133 Plus 2.0 Array in cartridge format
   - Strong signal and fold change correlation to previous plate and cartridge designs
• Multiple independent measurements per transcript for increased confidence in your results
   - Interrogates more sequence than a single probe measurement
• Conveniently packaged
   - Individual 16-, 24-, and 96-array plates available
   - All processing trays included
Content Profile:
Sequences used in the design of the arrays were selected from GenBank™, dbEST and RefSeq. The majority of sequence clusters were created from the UniGene database (Build 133, April 20, 2001) and refined by analysis and comparison with a number of other publicly available databases, including the Washington University EST trace repository and the University of California, Santa Cruz Golden-Path human genome database (April 2001 release).
An additional set of sequence clusters were created from Build 159 of UniGene (January 25, 2003) and refined by analysis and comparison with a number of other publicly available databases, including the Washington University EST trace repository and the NCBI human genome assembly (Build 31). Sequences were further analyzed for correct orientation, false priming, false clustering, alternative splicing and alternative polyadenylation.
*Two design changes were introduced with the GeneChip GeneChip HT HG-U133+ PM Array Plate:
• Only perfect match (PM) probes from the cartridge design were retained while mismatch (MM) probes were removed.
• Empirical data was used to select the best-performing probes resulting in reducing the number of PM probes
   - 42,461 probe sets were reduced from 11 to nine probes
   - Six probe sets were reduced from 11 to 10
For Research Use Only. Not for use in diagnostic procedures.
사양
제품라인GeneChip
수량1 kit
유형HT HG-U133+PM Array Plate
어레이Transcriptome Profiling
형식16-array Plate
어레이 수16 arrays
Human
Unit SizeEach
자주 묻는 질문(FAQ)
Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?
The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.
How long does it take to scan the plate?
96 arrays = 4-4.6 hours
24 arrays = 1.5-2 hours
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
If I have cartridges, can I move to PM only arrays in the middle of my project?
No, continue to finish products using the same procedures and products. New projects can be moved to PM only arrays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What QC metrics will I use to assess the quality of my data?
Mean absolute relative log expression: MA (RLE)
-Detection of 20X spikes, Bio B, Bio C, Bio D & Cre
-House keeping controls
-Poly A controls
-Antigenomic GC - 12 background probes
-PM mean
Please refer to the QC Metrics for Human, Mouse and Rat HT PM Array Plates QRC for more information (http://tools.thermofisher.com/content/sfs/manuals/ hmr_qc_metrics_qrc.pdf).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which algorithm do I use to run the analysis for these new HT PM Plate Arrays?
We recommend running the RMA algorithm in the Transcriptome Analysis Console (TAC).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.