PrimeView Human Genome U219 Array Plate, 1 x 96 array plate - FAQs
PrimeView Human Genome U219 Array Plate, 1 x 96 array plate - FAQs
View additional product information for PrimeView Human Genome U219 Array Plate - FAQs (901595, 901605, 901604)
2 product FAQs found
Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?
The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.
Are the PrimeView Human Genome U219 Array Strip and PrimeView Human Genome U219 Array Plate the same as the PrimeView Human Gene Expression Array?
Yes, the PrimeView Human U219 Array and the Primeview Human Gene Expression Array are indeed the same array (including strip and plate).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.