Axiom™ GeneTitan™ Consumables Kit - FAQs
Axiom™ GeneTitan™ Consumables Kit - FAQs
View additional product information for Axiom™ GeneTitan™ Consumables Kit - FAQs (901606)
2 product FAQs found
What are the dimensions of the wells of the hybridization and stain trays in the Axiom GeneTitan Consumables Kit (Cat. No. 901606)?
The wells of both the hybridization and the stain trays in the Axiom GeneTitan Consumables Kit have the same dimensions:
L x W x H: 0.8 cm x 0.8 cm x 0.8 cm
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the dimensions of the trays in the Axiom GeneTitan Consumables Kit (Cat. No. 901606)?
The dimensions of the trays in the Axiom GeneTitan Consumables Kit are as follows:
- Hybridization tray (L x W x H): 12.5 cm x 8.5 cm x 1.5 cm
- Scan tray (L x W x H): 12.5 cm x 8.5 cm x 1 cm
- Stain tray (L x W x H): 12.5 cm x 8.5 cm x 1 cm
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.