PrimeView™ Human Genome U219 Array Strip - FAQs
PrimeView™ Human Genome U219 Array Strip - FAQs
View additional product information for PrimeView™ Human Genome U219 Array Strip - FAQs (901613)
1 product FAQs found
Are the PrimeView Human Genome U219 Array Strip and PrimeView Human Genome U219 Array Plate the same as the PrimeView Human Gene Expression Array?
Yes, the PrimeView Human U219 Array and the Primeview Human Gene Expression Array are indeed the same array (including strip and plate).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.