GeneChip™ Porcine Gene 1.1 ST Array Strip - FAQs
GeneChip™ Porcine Gene 1.1 ST Array Strip - FAQs
View additional product information for GeneChip™ Porcine Gene 1.1 ST Array Strip - FAQs (901798)
1 product FAQs found
Does the GeneChip miRNA 3.1 Array Strip require a different hyb/wash/stain kit than the GeneChip 3' IVT arrays and GeneChip ST array strips?
The hyb/wash/stain kit for the GeneChip miRNA 3.1 Array Strips is different than that for the 3' IVT arrays and GeneChip ST array strips. Additional wash A steps have been added to ensure adequate washing after the array strips have been stained.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.