GeneChip™ Arabidopsis Gene 1.1 ST Array Plate - FAQs
GeneChip™ Arabidopsis Gene 1.1 ST Array Plate - FAQs
View additional product information for GeneChip™ Arabidopsis Gene 1.1 ST Array Plate - FAQs (901913)
30 product FAQs found
What is contained in the tab-delimited format of the GeneChip probe sequence download file?
The tab-delimited probe sequence file contains the following information:
-Probe Set Name
-Probe X: The X coordinate of the probe sequence on the GeneChip probe array.
-Probe Y: The Y coordinate of the probe sequence on the GeneChip probe array.
-Probe Interrogation Position: The base position on the consensus/exemplar sequence where the central base of the probe aligns, which is the 13th base of a 25mer probe.
-Probe Sequence: The 25-base perfect match sequence.
-Target Strandedness: The sense/antisense orientation of the target sequence that can hybridize with the probe sequence.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the NetAffx Analysis Center?
The NetAffx Analysis Center used to contain information and files regarding array content, probe sets, and functional annotations. However, the NetAffx Analysis Center has been retired. Much of the content provided by the NetAffx Analysis Center can now be accessed on the arrays’ thermofisher.com product pages or by contacting Technical Support (techsupport@thermofisher.com).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?
The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.
What is an Event Score in TAC 4.0 Software?
TAC 4.0 includes two algorithms for identifying alternative splicing events: the TAC 2.0 algorithm and the new EventPointer. Algorithmic determination of alternate splicing remains a challenging problem. TAC 4.0 supports two different approaches that have different sets of strengths and weaknesses. After considerable testing, the new TAC 4.0 “'Event Score” leverages both previous TAC 2.0 event estimation score and Event Pointer p-value and sorts the most likely alternative splicing events to the top. Of course, the TAC 2.0 event score and EventPointer p-values remain individually available.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the new software components of TAC 4.0?
LIMMA: LIMMA stands for Linear Models for MicroArray data. It is an R/Bioconductor software package that provides an integrated solution for analyzing data from gene expression experiments. It contains rich features for handling complex experimental designs and for information borrowing to overcome the problem of small sample sizes. Over the past decade, LIMMA has been a popular choice for gene discovery through differential expression analyses of microarray data. There are ˜8000 citations using LIMMA and Affymetrix arrays. The TAC 4.0 interface exposes the core differential expression analysis functionality including real covariates and random factors. In addition, the interface simplifies the creation of the design and contrast matrices that specify the experimental design and comparisons for the analysis.
Batch Effect Adjustment: Batch effects are systematic changes in microarray sample intensities that reflect changes in the assay sometimes found in different batches. These effects occur more commonly in larger studies in which all of the samples cannot be processed at the same time. TAC 4.0 enables the interface to the ComBat batch adjustment algorithm, which can remove the batch effects from the signals.
EventPointer: EventPointer is a Bioconductor package that identifies alternative splicing events in microarray data. TAC 4.0 incorporates an interface to this package.
Exploratory Grouping Analysis: Exploratory Grouping Analysis (EGA) is an interface to a set of R packages that offer the ability to examine the relationships between multiple microarray samples. While the scientist typically has a preconceived idea regarding the classification of the samples in an experiment, the resulting data often show additional substructure due to unexpected biological differences or batch effects. The EGA interface enables the identification of this substructure. Biological differences can be further explored using LIMMA differential expression analysis. Batch effects can be removed using ComBat to prevent them from obscuring the biology of interest.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
If I have TAC 3.1 .TAC files (TAC analysis files), can I load these into TAC 4.0 Software or will I need to reanalyze?
TAC 3.1 .TAC files cannot be opened in TAC 4.0 Software. Studies will need to be reprocessed in TAC 4.0. The new analysis can be run from .CEL files or .CHP files.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
In TAC 4.0 Software, can I measure the quality of a single hybridization without the rest of the experiment?
We do not recommend this. In large-scale expression experiments using similar sample types, researchers are likely to develop their own single-array guidelines on what metric values are predictive of high- or poor-quality samples. However, these guidelines are likely to be dependent on sample type and we are unable to recommend such guidelines for all possible situations. Note that the trend toward favoring model-based signal estimation algorithms (for all microarray experiments even beyond the Thermo Fisher platform) makes single-array quality determination very difficult due to the necessity of simultaneously analyzing multiple arrays to calculate signal estimates.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
In the QC metrics for TAC 4.0 Software, one of my samples has skewed values for several of the metrics, but the values aren't grossly out of line. I could make a case to keep it in or discard it. What should I do?
If a sample appears “on the line”, it is best to leave the sample in the experiment for analysis and simply flag it as questionable. During the initial analysis, treat all samples uniformly. Once candidate genes have been identified, review how the questionable sample changes data relative to the other replicates within the sample. It is always possible to remove a sample later in the analysis workflows.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which quality metric is most predictive of the quality of hybridization when viewing the QC results in TAC 4.0 Software?
Pos_vs_Neg_AUC is a good first-pass metric. A value below 0.7 is an indicator that sample problems may exist. However, a value greater than 0.7 does not guarantee that the sample is good.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are there any circumstances in which I would benefit from using RMA over SST-RMA with whole transcriptome arrays?
Users will benefit from using the SST-RMA when comparing data with those processed using the RMA summarization only. RMA is the suggested analysis pipeline to use when doing QC on FFPE or degraded samples. For downstream analysis of FFPE or degraded samples, SST-RMA is the suggested analysis pipeline to use.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can I expect to see with Signal Space Transformation-Robust Multi-Chip Analysis (SST-RMA) compared to RMA algorithm?
When filtering by fold-change cutoffs, expect a larger number of differentially expressed genes compared to RMA. When comparing the number of differentially expressed genes (defined by fold-change filters) to other methods, such as RT-PCR and RNA-Seq, expect this number to align more with these methods when using SST-RMA. There is no impact on sensitivity or specificity of data. SST-RMA was designed to address comparability to other technologies. The same experimental design recommendations still apply when designing an expression study with microarrays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the Signal Space Transformation-Robust Multi-Chip Analysis (SST-RMA) algorithm?
Historically, microarrays were perceived to underestimate fold-change values when compared to other methods such as RT-PCR. For customers who are filtering using fold-change cutoff, SST-RMA addresses this “fold-change compression” by applying a GC correction and also by transforming the microarray data signal to a similar signal space of other methods in a pre-processing step prior to RMA. No changes have been made to RMA. SST-RMA is the default algorithm for Human Transcriptome Arrays and Clariom D Human, Mouse, and Rat assays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Where are the TAC 4.0 library files located on the website?
Library files are on each array type product page. Alternatively, library files can be installed directly through TAC 4.0 by going to the 'Preferences' tab and clicking the 'Download Library Files' button located at the top left (NetAffx account required).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I install TAC 4.0 Software on my AGCC workstation computer that controls the Affymetrix hardware?
Due to the amount of memory that TAC needs to operate, we STRONGLY recommend that you DO NOT install TAC 4.0 on production AGCC computers being used for scanning and operating fluidics systems.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What files are required for analysis in TAC 4.0 Software?
Both .CEL and .CHP files are accepted in TAC 4.0 Software. The exception will be previous *.exon*. CHP files and *.alt-splice*. CHP files. These two types of CHP files are no longer supported in TAC 4.0. .ARR files are recommended as they contain sample attribute information. The .CHP file is generated by TAC 4.0 in the normalization and summarization process.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the new features of TAC 4.0 Software?
TAC 4.0 is a new software that combines many of the features of Expression Console 1.4 with the biological analysis capabilities of TAC into one workflow. Some of the new features include:
- Perform QC and statistical analysis in one software
- Ability to analyze 1000 samples
- Integration of LIMMA package (LInear Modeling for MicroArrays package) into TAC 4.0 to analyze complex data
- Perform batch effect adjustment (new)
- Perform repeated measure analysis (improved in TAC 4.0)
- Perform random factor analysis (new)
- Perform real covariate analysis (new)
- Ability to perform Exploratory Grouping Analysis (EGA)
- Integration of Event Pointer algorithm for Alternative Splicing Analysis
- Improved visualization with Splicing Viewer
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the minimum "computer requirements" for TAC 4.0 Software?
Expression array CEL files are processed and analyzed in TAC 4.0 Software. This is a free download from the Thermo Fisher website for processing CEL files.
Here are the minimum requirements:
Microsoft Windows 7 and 10 (64 bit) Professional
CPU: 2.83 GHz
Memory: 8 GB RAM
Hard drive: 150 GB free hard disk space
Web browser: Internet Explorer 11 or greater and Microsoft Edge
Here is the recommended hardware:
Microsoft Windows 7 and 10 (64 bit) Professional
CPU: Intel Pentium 4X 2.83 GHz processor (Quad Core)
Memory: 16 GB RAM
Hard drive: 150 GB free hard disk space
Web browser: Internet Explorer 11 or greater and Microsoft Edge
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When should data analyzed with RMA be re-analyzed using SST-RMA?
You should re-analyze your data if comparing with RT-PCR or RNA-Seq data using fold change values for filtering.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Two alternative splicing detection methods were described in the white papers on your web site. Are they related to the ANOSVA SVD algorithm that was published in Bioinformatics in 2005 by Cline, et al?
No, the methods in the white papers are not ANOSVA which was published in the paper referenced (http://bioinformatics.oxfordjournals.org/cgi/content/abstract/21/suppl_1/i107). One note about ANOSVA is that it was developed on a combination exon/junction array. Some preliminary work comparing ANOSVA and PAC (one of the two methods described in the white papers) on the exon array tissue panel suggests that the MiDAS and the robust PAC methods presented in the white papers are a better way to go.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do you move the .cel files out of GCOS to be imported into Expression Console?
GCOS users must use DTT v1.1, using the Flat File option, to transfer files to be analyzed by the Expression Console software from the GCOS database to an independent folder.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When I loaded the GeneChip Exon Array design information and array data into IGB, it seems that the probes were selected from outside of the RefSeq exons. Why?
The most likely explanation is that a different version of the genome assembly has been used to display the array design information and the array results. At launch, two versions of the library files are provided for array analysis corresponding to the Human Genome Build 34 and 35. Take care to use a consistent version number to match the array design with the actual array data for visualization in IGB.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do I correlate the SNPs on the mapping GeneChip array with this design?
Associations between SNPs and exon probe sets can be obtained by using genome assembly position information which is provided for both the mapping array and the exon array. One useful tool for doing this is the UCSC Table Browser at http://genome.ucsc.edu/cgi-bin/hgTables.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I get gene-level expression results for GeneChip Exon arrays?
The current exon array analysis software allows for the aggregation of multiple exon level probe sets into a larger "meta probe set". PLIER signal estimates and DABG detection values are then computed for these meta probe sets. The definition and grouping of the exons into a gene can have a significant impact on the final signal value of a particular gene. Affymetrix recommends using the "core gene" grouping or the "full gene" grouping files to derive the gene-level signal that should most resemble the expression of the constitutive exons.
See the "Gene Signal Estimates from Exon Arrays" (https://tools.thermofisher.com/content/sfs/brochures/exon_gene_signal_estimate_whitepaper.pdf) white paper for more information.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is DABG, PLIER?
DABG stands for "detection above background" and is a detection metric generated by comparing Perfect Match probes to a distribution of background probes. This comparison yields a p-value which is then combined into a probe set level p-value using the Fischer equation. PLIER stands for "Probe Logarithmic Intensity Error" and is a model-based signal estimator which benefits from multi-array analysis.
For more information on DABG, see the
"Exon Array Background Correction" (https://tools.thermofisher.com/content/sfs/brochures/exon_background_correction_whitepaper.pdf) white paper; for more information on PLIER, refer to the PLIER Technical Note. (https://tools.thermofisher.com/content/sfs/brochures/plier_technote.pdf)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can I do to reduce the analysis time required to generate probe set level signal when working with GeneChip Exon Arrays?
Two primary factors can affect the length of analysis time: 1) the number of arrays analyzed at one time, and 2) the number of probe sets to be included in the analysis. Reducing the size of either one of these two factors will increase the analysis speed.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long will it take for me to analyze the data for the GeneChip Exon Arrays?
Normalization of .cel files takes 1-3 minutes and generation of DABG and PLIER probe set level summaries takes about 40 minutes per .cel file. Further downstream analysis will depend on which data analysis techniques are being applied.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What analysis software packages are available for GeneChip Exon Array data analysis?
GeneChip Command Console (AGCC) is provided to control the fluidics station, 7G Scanner and generate .dat and .cel files. Probe level analysis of the .cel files is carried out by Expression Console. This software allows for the generation of signal estimates and detection p-values at the probe set level for either exon-level or gene-level analysis. Expression Console Software is freely downloadable from our web site and supported by the Technical Support team.
For additional higher levels of analysis, such as alternative splicing detection, a few experimental algorithms are published as methods in respective white papers. Users will need to implement these methods in advanced statistical analysis software packages. These methods have been developed based on experience with limited sample data sets, and further fine-tuning may be required depending on the user's unique biological systems. It is anticipated that new methods will continue to emerge to better support Exon Array analysis in the near future.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I compare the Signal value between two exons directly to obtain a ratio of two different transcripts for your GeneChip Exon Arrays?
No. Like probes on other current expression arrays, there are probe-specific effects that cause each probe set to respond differently to the target abundance. Thus, the comparison of signal ratios between different probe sets to compare their expression levels is invalid.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the basic quality assessment metrics for the GeneChip Exon Array?
The standard Affymetrix expression control sets (i.e., bacterial spikes) including both the hybridization control spikes and poly-A RNA control spikes are present on the Human Exon 1.0 ST Array and can be used to assess quality in a qualitative manner as used previously. As a new feature unique for the Exon Array, the Expression Console software supports the extraction of residuals from the PLIER model when running a multiple-array analysis. Such residuals can be used to identify outlier arrays and poor performing probes.
In addition, for most of the 100 normalization control genes used in the design of the GeneChip Human Genome U133 Array, probe sets representing both their intron and exon regions have been tiled on the Exon Array. The exon and intron probe set metrics (i.e., % detection p-value <= 0.05, mean/median signal) can be used as an additional method to identify problematic outlier arrays or analysis problems.
For these new experimental analysis metrics, there are no simple standard numeric threshold values that we can recommend at this moment as cutoffs for identifying good-performing or poor-performing arrays. However, it has been found that they are valuable when comparing relative values across a set of experiments for identifying outlier arrays. See the "Quality Assessment of Exon Arrays" (https://tools.thermofisher.com/content/sfs/brochures/exon_gene_arrays_qa_whitepaper.pdf) white paper for more information.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How big are the .dat and .cel GeneChip Exon Array files?
The .dat files are approximately 750 MB in size, and the binary .cel files are approximately 60 MB in size.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.