Sometimes, there are transcript clusters in the GeneChip Human Transcriptome Array (HTA) 2.0 with multiple associated gene symbols, and it appears that the probes in the cluster span multiple adjacent/overlapping genes.
For example: The transcript TC04000948.hg.1 is associated with the gene symbols ABCA11P//ZNF721 in the GeneChip HTA 2.0 annotation file. This is one of those infrequent cases where multiple genes will co-exist in the same location. In this particular case, one of them is a zinc-finger protein, and the other one is a pseudogene. Pseudogenes are often defined as "not having any known function" or "not expressed". However, as the knowledge of biology progresses, the definition of "function" changes and it may be found that these do have function, just that it is not known. Pseudogenes occasionally are transcribed, and sometimes, they are just artifacts of ancient or recent duplication events.
Find additional tips, troubleshooting help, and resources within our
Microarray Analysis Support Center.