Axiom™ Biobank Genotyping Array, 96 array plate
Applied Biosystems™

Axiom™ Biobank Genotyping Array, 96 array plate

The Axiom™ Biobank Genotyping Array was designed by and for thought leaders in the human genetics community for high-throughput, high-valueRead more
Have Questions?
Catalog NumberQuantity
9021861 plate
Catalog number 902186
Price (MXN)
-
Quantity:
1 plate
The Axiom™ Biobank Genotyping Array was designed by and for thought leaders in the human genetics community for high-throughput, high-value genotyping of large sample cohorts to explore the genetics of complex diseases and translational research with a single comprehensive low-cost solution. This array contains one 96-array plate that allows for genome-wide genotyping of large sample collections such as those screened at biobanks, genome centers, and core labs.

GWAS
• Intelligent marker selection enables imputation of millions of additional SNPs
• The genomic content module of each array is optimized for specific populations and is customizable to any 1000 Genomes population study by adding additional markers of your choosing

• Immune function, including content covering the major histocompatibility complex (MHC), human leukocyte antigen (HLA), and killer-cell immunoglobulin-like receptors (KIR)

Pharmacogenomic markers
• Markers were selected to represent phases of absorption, distribution, metabolism, and excretion (ADME)

Inflammation and HLA
• Contains markers with evidence for association with autoimmune and inflammation
• Covers variants in genes in the HLA and KIR regions known to be important in immune response

Exome
• Rare, non-synonymous coding SNPs and indels in protein coding regions of the genome
• A majority of variants are rare with minor allele frequency (MAF) <1%
• Newly discovered loss-of-function content
• SNPs and indels identified from a sequencing initiative of 26,000 individuals
• Known disease-causing mutations

eQTLs
• Markers that have known associations to RNA expression traits
• Captures unique eQTLs contained in the NCBI Genotype-Tissue Expression (GTEx) eQTL database, GEUVIDAS project, and several other discovery projects

Human disease
• Blood phenotypes
• Cancer common variants
• Cardiometabolic

Choose your content
• Order the pre-designed array and customize it with markers of your choice
• Custom markers may be de novo or selected from the Axiom Genomic Database of genotype-tested markers

Please note: Reagent kits do not include plastic consumables required to run the assay on the Beckman Biomek™ FXP Target Prep Express System. Please contact us for a list of required plastic consumables.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
FormLiquid
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesHuman
Unit SizeEach

Frequently asked questions (FAQs)

For the Axiom Biobank Genotyping Array, 96 array plate (Cat. No. 902186), which instrument is required to run the assay?

The Beckman Biomek™ FXP Target Prep Express System is required for running the Axiom Biobank Genotyping Array, 96 array plate (Cat. No. 902186).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?

The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.