GeneChip™ Human Transcriptome Array 2.0, 2 arrays - FAQs
GeneChip™ Human Transcriptome Array 2.0, 2 arrays - FAQs
View additional product information for GeneChip™ Human Transcriptome Array 2.0 - FAQs (902162, 902233)
5 product FAQs found
Does the GeneChip Human Transcriptome Array 2.0 (Cat. No. 902233, 902162) detect gene fusion?
The GeneChip Human Transcriptome Array 2.0 does not detect gene fusion.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How long does it take for the GeneChip Human Transcriptome Array 2.0 (Cat. No. 902162) to scan?
The GeneChip Human Transcriptome Assay 2.0 takes approximately 35 min to scan.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why are multiple gene symbols associated with the same Transcript Cluster IDs in GeneChip Human Transcriptome Array 2.0?
Sometimes, there are transcript clusters in the GeneChip Human Transcriptome Array (HTA) 2.0 with multiple associated gene symbols, and it appears that the probes in the cluster span multiple adjacent/overlapping genes.
For example: The transcript TC04000948.hg.1 is associated with the gene symbols ABCA11P//ZNF721 in the GeneChip HTA 2.0 annotation file. This is one of those infrequent cases where multiple genes will co-exist in the same location. In this particular case, one of them is a zinc-finger protein, and the other one is a pseudogene. Pseudogenes are often defined as "not having any known function" or "not expressed". However, as the knowledge of biology progresses, the definition of "function" changes and it may be found that these do have function, just that it is not known. Pseudogenes occasionally are transcribed, and sometimes, they are just artifacts of ancient or recent duplication events.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the difference between GeneChip Human Transcriptome Array v1(HTAv1) and GeneChip Human Transcriptome Array 2.0 (HTA 2.0)?
HTAv1, also known as Glue Grant Human Transcriptome Array (GG-H), is a custom array made in collaboration with Stanford Genome Technology, Wing Wong's lab at Stanford, and the Inflammation and Host Response to Injury program (“Glue Grant”). The array was designed to interrogate gene expression, alternative splicing, detection of coding SNPs and non-coding transcription. Permissions from the group are required to use this array. HTA 2.0 is one of our catalog arrays. This array follows a similar design as the HTAv1 but focus on the whole transcript gene expression and alternative splicing of coding and non-coding transcripts. We provide assay and software for the processing and analysis of this array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.