Product Image
Applied Biosystems™

Axiom™ 384HT GeneTitan™ Consumables Kit

Have Questions?
Catalog NumberQuantity
9022341 kit
Catalog number 902234
Price (USD)
-
Quantity:
1 kit
The Axiom™ GeneTitan™Consumables Kit is used with the Axiom Wheat Breeder's Genotyping Array (384 format), the Axiom Trout Genotyping Array (384 format), or the Axiom 384HT myDesign™ Custom Array (each sold separately).

Includes consumables required to process one Axiom 384-Array Plate on the GeneTitan Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
For Use With (Equipment)GeneTitan Multi-Channel (MC) Instrument
Quantity1 kit
TypeConsumables Kit
Unit SizeEach
Contents & Storage
Store all at room temperature

Contains:
5 x 384 layout stain trays
1 x 384 layout Hybridization tray
1 x 384 layout Scan tray
6 x 384 layout Scan and Stain tray covers