GeneChip™ miRNA 4.0 Array, 30 arrays - FAQs
View additional product information for GeneChip™ miRNA 4.0 Array - FAQs (902411, 902412, 902413)
19 product FAQs found
Where can I find the species information for your GeneChip miRNA arrays?
The species information can be found in the annotation file of the miRNA array of interest.
Please complete the following steps:
- Go to www.thermofisher.com and go to the product page of the array of interest.
- Scroll down to support files.
- Download the annotation files for the miRNA array of interest, and open in excel.
- In the file, there will be a column, species scientific name. This column will have all the species present on the array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For GeneChip miRNA arrays, are External RNA Controls Consortium (ERCC) controls tiled on the array?
The ERCC controls are not tiled onto GeneChip miRNA arrays because they would not work with the miRNA array/assay system. The FlashTag Biotin HSR RNA Labeling Kits used to label miRNAs is optimized for shortRNAs. Even though longer mRNAs like the ERCC controls will be labeled, the HWS (hyb/wash/stain) conditions are not optimal and consequently, the longer mRNAs typically do not get detected.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which of the gene arrays are compatible with the miRNA interaction network in Transcriptome Analysis Console (TAC) Software?
The miRNA interaction network in TAC Software is available for all gene arrays, including model organisms. To have the interaction network available, the same samples must have been processed through both the gene array and the miRNA array. Additionally, the gene array being used must have a gene symbol column displayed in their annotation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Does GeneChip miRNA 4.0 Array contain any feline miRNA content?
No, GeneChip miRNA 4.0 Array does not contain any microRNAs specific to felines.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How are sno/sca and hairpin probe sets selected and how does that impact my signal summarization?
Probes for snoRNA, scaRNA, and hairpins are selected to maximize probe response to target concentration in the sample while minimizing cross-hybridization to other potential targets in the sample. In order to minimize cross-hybridization, potential targets are inferred from the landscape of known sequences from the input dataset and used in a filtering process called pruning which penalizes probe candidates that may cross-hybridize to unwanted targets. As the landscape of known sequences improves, we can improve our pruning set to avoid probe candidates previously thought to be unique. An improved pruning set will reduce the chances a probe will cross-hybridize to unwanted target, improving the probe set representation of the intended target. As a result of the improved probe selection, few probe sets are required to represent the same number of sequences.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I combine the CEL files from GeneChip miRNA Arrays, miRNA Array Strips, and miRNA Array Plates into the same experiment?
No, the CEL files from GeneChip miRNA Array, Thermo Fisher Scientific miRNA Array Strip, and miRNA Array Plates are not compatible.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the difference between GeneChip miRNA Array, miRNA Array Strip, and miRNA Array Plate?
The only difference is the form factor of the array. The arrays have the exact same design and contain the same number of probes and probe sets.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the orientation of the probes on the Thermo Fisher Scientific miRNA arrays?
Probes on the array detect sense target.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the recommended hybridization time for Thermo Fisher Scientific miRNA Arrays?
GeneChip miRNA Array 17 +/- 1 hr
Thermo Fisher Scientific miRNA Array Plates 17 +/- 1 hr
Thermo Fisher Scientific miRNA Array Strips 20 +/- 1 hr
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the difference between GeneChip miRNA 4.0 Cartridge Array, miRNA 4.1 Array Strip, and Affymetrix miRNA 4.1 Array Plate?
The arrays have the exact same design and contain the same number of probes and probe sets.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
I would like to check my miRNA Array data with qRT-PCR. In addition to my miRs of interest, what other qRT-PCR controls should I run?
At least 5 miRs or SnoRNAs should be used to normalize:
RU44, RU48, and/or U6 microRNAs that are not changed among your samples, and are at least 5X over background, according to your microarrays. These miRs might include miR 15,16, 17, or let 7a, let 7b, let7c
All 5 (or more) of these RNAs should not show any change among the samples. Average some or all of these to get the normalization factor, and apply to your qRT-PCR data.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can you explain the correlation of miRNA Array data to miRNA qPCR data?
qPCR is not yet the gold standard for miRNA validation. Unlike mRNA validation, in which the amplicon is already present in the sample, miRNA qPCR requires the amplicon to be synthesized by combining the sample with either a specially designed hairpin molecule, adding a 3' polyA tail, or some other manipulation of the microRNA sample. The amplicon-building process will be different from sample to sample and will result in variability in the PCR results. However, it is still very important to validate the array results with another method, like PCR. The trends in up and down regulation should match in direction, even if they do not match in magnitude.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are there any articles that describe miRNA Array data analysis?
F Sato. Intra-Platform Repeatability and Inter-Platform Comparibility of microRNA microarray technology. PLoS ONE May 2009, volume 4, issue 5, e5540.
D Sarkar. Quality Assessment and data analysis for microRNA expression arrays. Nucleic Acids Research 2009, vol. 37, no. 2, e17 (doi: 10.1093/ner/gkn932).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Should I disregard all array data for probes that are not the same species as my RNA sample?
Most researchers eliminate the irrelevant species from analysis, because the extra data can be cumbersome. However, looking at multi-species probes may provide an increased confidence in the data. We typically observe a clustering of cross-species miRs that are identical, or nearly identical.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the recommended OD 260:280 ratios for input RNA for downstream array analysis with miRNA GeneChip microarrays?
We do not have a specific recommendation. A high-quality RNA sample will have a ratio of at least 1.95. However, if microRNAs are present in samples with lower 260:280 ratios, these samples can still be used.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Should I treat my RNA sample with DNAse for FlashTag labeling?
DNAse treatment is optional. It is not necessary for RNA samples that have trace amounts of genomic DNA contamination, but it may be beneficial for RNA samples that are highly contaminated with genomic DNA, to more accurately quantitate the RNA. After treating your RNA with DNAse, it is essential that the DNase be inactivated completely before proceeding with the FlashTag procedure, to prevent degradation of the FlashTag reagent (Vial 5). A variety of RNA purification columns/kits may be used to inactivate the DNase. Inactivation of the DNase by high temperature may not completely inactivate the enzyme.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How should I quantitate my RNA for downstream miRNA array analysis?
To accurately determine the concentration of the RNA sample, we recommend Quant-iT RiboGreen RNA Assay Kit (Cat. No. R11490) or the NanoDrop ND-1000 Spectrophotometer.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Should I enrich my total RNA for miRNA for downstream miRNA array analysis?
Either total RNA or low melecular weight (LMW) RNA can be labeled with FlashTag Biotin HSR. Using total RNA can save time and money, and prevent sample loss.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How should I isolate/purify my RNA for miRNA array analysis?
Any kit for purification of total RNA or LMW (Low Molecular Weight) RNA will be compatible with FlashTag Biotin HSR. Elute or resuspend the RNA in nuclease-free water. Ensure that the purification method retains low molecular weight species. Some commercial products that have been tested successfully with FlashTag Biotin HSR include:
mirVana miRNA Isolation Kit
RecoverAll Total Nucleic Acid Isolation Kit for FFPE
PureLink miRNA Isolation Kit
TRIzol reagent (total RNA only) with additional overnight -20 degrees C precipitation step during isopropanol precipitation.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.