GeneChip™ Rat Gene 2.0 ST Assay, 30 samples - FAQs
GeneChip™ Rat Gene 2.0 ST Assay, 30 samples - FAQs
View additional product information for GeneChip™ Rat Gene 2.0 ST Assay - FAQs (902466, 902467)
2 product FAQs found
Are pseudogene databases included in the design of expression arrays?
Pseudogene databases were not included in the design of expression arrays.
How long can I store labeled cDNA when working with expression microarrays?
Labeled material can be stored for 2 weeks at -20 degrees C.