GeneChip™ Mouse Gene 2.0 ST Array, 2 arrays - FAQs
GeneChip™ Mouse Gene 2.0 ST Array, 2 arrays - FAQs
View additional product information for GeneChip™ Mouse Gene 2.0 ST Array - FAQs (902500, 902118, 902119)
3 product FAQs found
Are pseudogene databases included in the design of expression arrays?
Pseudogene databases were not included in the design of expression arrays.
How long can I store labeled cDNA when working with expression microarrays?
Labeled material can be stored for 2 weeks at -20 degrees C.