Clariom™ D Assay, mouse, 30 reactions - FAQs
Clariom™ D Assay, mouse, 30 reactions - FAQs
View additional product information for Clariom™ D Assay, mouse - FAQs (902513, 902514)
26 product FAQs found
Why is it important to use Tough-Spots® labels when using GeneChip cartridge arrays?
Tough-Spots® labels are small adhesive stickers used to temporarily seal the backs of cartridge arrays during the overnight hybridization step. They are required to prevent loss of volume due to evaporation through the septa. We recommend using Tough-Spots® labels on Rolls from USA Scientific (Item No. 9185-0000)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the respective hybridization volumes based on array format for Affymetrix gene expression arrays?
Proper hybridization volume is critical to obtaining an even signal across a given array. Too little volume can lead to black circles in the middle of the array. Too much volume can leak out of the back of the array. The correct hybridization volume leaves enough room for a small air bubble to circulate around the array surface during the overnight hybridization. Here are the recommended hybridization and fill volumes based on the array format:
Array Format; Hybridization Volume; Fill Volume
- 49 Format (Standard); 200 µL; 250 µL
- 64 Format; 200 µL; 250 µL
- 100 Format (Midi); 130 µL; 160 µL
- 169 Format (Mini); 80 µL; 100 µL
- 400 Format (Micro); 80 µL; 100µL
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why is strand-specificity important when performing Clariom D and Clariom S assays?
Clariom D arrays have probes that cover all known regions of transcription including probes in overlapping regions from both strands. To obtain strand-specific information from the Clariom D arrays, the WT Pico and WT Plus reagents (which are strand-specific) must be used. This is important because without strand-specific reagent, it would not be possible to decipher the source strand of DNA, which makes it challenging to untangle true gene- and exon- level expression and alternative splicing events.
Strand-specificity is significantly less important for customers interested in gene-level only information (i.e., those using Clariom S) as compared to customers who want to understand the complexities of the whole transcriptome including identifying antisense transcripts and ncRNA (i.e., those using Clariom D). While strand-specificity is less important for gene-level expression only, probes within regions of overlapping transcription from both strands are avoided in the Clariom S array design (unlike Clariom D). This is important because if Clariom S did not preserve strand-specificity, there could be an overestimation of gene-level expression causing false positive or negative results. With Clariom S having a "stranded" design, it does not necessarily need a strand-specific reagent kit.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is an Event Score in TAC 4.0 Software?
TAC 4.0 includes two algorithms for identifying alternative splicing events: the TAC 2.0 algorithm and the new EventPointer. Algorithmic determination of alternate splicing remains a challenging problem. TAC 4.0 supports two different approaches that have different sets of strengths and weaknesses. After considerable testing, the new TAC 4.0 “'Event Score” leverages both previous TAC 2.0 event estimation score and Event Pointer p-value and sorts the most likely alternative splicing events to the top. Of course, the TAC 2.0 event score and EventPointer p-values remain individually available.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the new software components of TAC 4.0?
LIMMA: LIMMA stands for Linear Models for MicroArray data. It is an R/Bioconductor software package that provides an integrated solution for analyzing data from gene expression experiments. It contains rich features for handling complex experimental designs and for information borrowing to overcome the problem of small sample sizes. Over the past decade, LIMMA has been a popular choice for gene discovery through differential expression analyses of microarray data. There are ˜8000 citations using LIMMA and Affymetrix arrays. The TAC 4.0 interface exposes the core differential expression analysis functionality including real covariates and random factors. In addition, the interface simplifies the creation of the design and contrast matrices that specify the experimental design and comparisons for the analysis.
Batch Effect Adjustment: Batch effects are systematic changes in microarray sample intensities that reflect changes in the assay sometimes found in different batches. These effects occur more commonly in larger studies in which all of the samples cannot be processed at the same time. TAC 4.0 enables the interface to the ComBat batch adjustment algorithm, which can remove the batch effects from the signals.
EventPointer: EventPointer is a Bioconductor package that identifies alternative splicing events in microarray data. TAC 4.0 incorporates an interface to this package.
Exploratory Grouping Analysis: Exploratory Grouping Analysis (EGA) is an interface to a set of R packages that offer the ability to examine the relationships between multiple microarray samples. While the scientist typically has a preconceived idea regarding the classification of the samples in an experiment, the resulting data often show additional substructure due to unexpected biological differences or batch effects. The EGA interface enables the identification of this substructure. Biological differences can be further explored using LIMMA differential expression analysis. Batch effects can be removed using ComBat to prevent them from obscuring the biology of interest.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
If I have TAC 3.1 .TAC files (TAC analysis files), can I load these into TAC 4.0 Software or will I need to reanalyze?
TAC 3.1 .TAC files cannot be opened in TAC 4.0 Software. Studies will need to be reprocessed in TAC 4.0. The new analysis can be run from .CEL files or .CHP files.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
In TAC 4.0 Software, can I measure the quality of a single hybridization without the rest of the experiment?
We do not recommend this. In large-scale expression experiments using similar sample types, researchers are likely to develop their own single-array guidelines on what metric values are predictive of high- or poor-quality samples. However, these guidelines are likely to be dependent on sample type and we are unable to recommend such guidelines for all possible situations. Note that the trend toward favoring model-based signal estimation algorithms (for all microarray experiments even beyond the Thermo Fisher platform) makes single-array quality determination very difficult due to the necessity of simultaneously analyzing multiple arrays to calculate signal estimates.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
In the QC metrics for TAC 4.0 Software, one of my samples has skewed values for several of the metrics, but the values aren't grossly out of line. I could make a case to keep it in or discard it. What should I do?
If a sample appears “on the line”, it is best to leave the sample in the experiment for analysis and simply flag it as questionable. During the initial analysis, treat all samples uniformly. Once candidate genes have been identified, review how the questionable sample changes data relative to the other replicates within the sample. It is always possible to remove a sample later in the analysis workflows.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which quality metric is most predictive of the quality of hybridization when viewing the QC results in TAC 4.0 Software?
Pos_vs_Neg_AUC is a good first-pass metric. A value below 0.7 is an indicator that sample problems may exist. However, a value greater than 0.7 does not guarantee that the sample is good.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Are there any circumstances in which I would benefit from using RMA over SST-RMA with whole transcriptome arrays?
Users will benefit from using the SST-RMA when comparing data with those processed using the RMA summarization only. RMA is the suggested analysis pipeline to use when doing QC on FFPE or degraded samples. For downstream analysis of FFPE or degraded samples, SST-RMA is the suggested analysis pipeline to use.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What can I expect to see with Signal Space Transformation-Robust Multi-Chip Analysis (SST-RMA) compared to RMA algorithm?
When filtering by fold-change cutoffs, expect a larger number of differentially expressed genes compared to RMA. When comparing the number of differentially expressed genes (defined by fold-change filters) to other methods, such as RT-PCR and RNA-Seq, expect this number to align more with these methods when using SST-RMA. There is no impact on sensitivity or specificity of data. SST-RMA was designed to address comparability to other technologies. The same experimental design recommendations still apply when designing an expression study with microarrays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the Signal Space Transformation-Robust Multi-Chip Analysis (SST-RMA) algorithm?
Historically, microarrays were perceived to underestimate fold-change values when compared to other methods such as RT-PCR. For customers who are filtering using fold-change cutoff, SST-RMA addresses this “fold-change compression” by applying a GC correction and also by transforming the microarray data signal to a similar signal space of other methods in a pre-processing step prior to RMA. No changes have been made to RMA. SST-RMA is the default algorithm for Human Transcriptome Arrays and Clariom D Human, Mouse, and Rat assays.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Where are the TAC 4.0 library files located on the website?
Library files are on each array type product page. Alternatively, library files can be installed directly through TAC 4.0 by going to the 'Preferences' tab and clicking the 'Download Library Files' button located at the top left (NetAffx account required).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I install TAC 4.0 Software on my AGCC workstation computer that controls the Affymetrix hardware?
Due to the amount of memory that TAC needs to operate, we STRONGLY recommend that you DO NOT install TAC 4.0 on production AGCC computers being used for scanning and operating fluidics systems.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What files are required for analysis in TAC 4.0 Software?
Both .CEL and .CHP files are accepted in TAC 4.0 Software. The exception will be previous *.exon*. CHP files and *.alt-splice*. CHP files. These two types of CHP files are no longer supported in TAC 4.0. .ARR files are recommended as they contain sample attribute information. The .CHP file is generated by TAC 4.0 in the normalization and summarization process.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the new features of TAC 4.0 Software?
TAC 4.0 is a new software that combines many of the features of Expression Console 1.4 with the biological analysis capabilities of TAC into one workflow. Some of the new features include:
- Perform QC and statistical analysis in one software
- Ability to analyze 1000 samples
- Integration of LIMMA package (LInear Modeling for MicroArrays package) into TAC 4.0 to analyze complex data
- Perform batch effect adjustment (new)
- Perform repeated measure analysis (improved in TAC 4.0)
- Perform random factor analysis (new)
- Perform real covariate analysis (new)
- Ability to perform Exploratory Grouping Analysis (EGA)
- Integration of Event Pointer algorithm for Alternative Splicing Analysis
- Improved visualization with Splicing Viewer
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the minimum "computer requirements" for TAC 4.0 Software?
Expression array CEL files are processed and analyzed in TAC 4.0 Software. This is a free download from the Thermo Fisher website for processing CEL files.
Here are the minimum requirements:
Microsoft Windows 7 and 10 (64 bit) Professional
CPU: 2.83 GHz
Memory: 8 GB RAM
Hard drive: 150 GB free hard disk space
Web browser: Internet Explorer 11 or greater and Microsoft Edge
Here is the recommended hardware:
Microsoft Windows 7 and 10 (64 bit) Professional
CPU: Intel Pentium 4X 2.83 GHz processor (Quad Core)
Memory: 16 GB RAM
Hard drive: 150 GB free hard disk space
Web browser: Internet Explorer 11 or greater and Microsoft Edge
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do I install Expression Console Software and TAC Software library files for human, mouse, or rat transcriptome assays?
Analysis library files and annotation files can be downloaded from within the TAC Software. Please refer to the respective user manuals for detailed instructions.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What software is available for analyzing the results from my human, mouse, or rat transcriptome assays?
Probe cell intensity data (CEL) from human, mouse, or rat transcriptome assays are analyzed in Transcriptome Analysis Console(TAC) Software. The application uses a version of RMA analysis to create CHP files. Transcriptome Analysis  Console Software can be downloaded at no charge from the website. Further statistical analysis may be performed in Affymetrix' Transcriptome Analysis Console (TAC) Software to obtain a list of differentially expressed genes and alternative splicing events. TAC Software also provides the visualization of genes, exons, junctions, transcript isoforms, pathways, and mRNA-miRNA regulatory networks.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How do I install AGCC library files for the human, mouse, or rat transcriptome arrays?
To install library files for the human, mouse, or rat transcriptome arrays for AGCC, simply run the executable library file installer.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What format are the human, mouse, or rat transcriptome arrays?
Human, mouse, or rat transcriptome arrays are 49-format.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which fluidics protocol should I use when processing human, mouse, or rat transcriptome assays?
FS450_0001 should be used when processing human, mouse, or rat transcriptome assays. Up-to-date fluidics scripts can be obtained from the website.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Can I analyze the results from my human, mouse, or rat transcriptome assays using GeneChip Operating Software (GCOS) or GeneChip Command Console Software (AGCC)?
Human, mouse, or rat transcriptome assays utilize an updated installer package that is not compatible with GCOS. Human, mouse, or rat transcriptome assays require AGCC for fluidics and scanning. The Expression Console Software is required to perform QC analysis of the human, mouse, or rat transcriptome array data. Expression Console Software can be downloaded at no charge from the website.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Which protocol should I use to prepare my sample for hybridization to GeneChip human, mouse, or rat transcriptome assays?
Affymetrix human, mouse, and rat transcriptome assays are available in three formats based on sample type and amount of input RNA:
Human, mouse, and rat transcriptome Pico assays compatible with 100-2,000 pg of RNA isolated from fresh, frozen, blood, or FFPE samples
Human, mouse, and rat transcriptome FFPE assays compatible with 20-200 ng of RNA isolated from FFPE samples
Human, mouse, and rat transcriptome assays compatible with 50-500 ng of RNA isolated from fresh, frozen, or blood samples
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
When should data analyzed with RMA be re-analyzed using SST-RMA?
You should re-analyze your data if comparing with RT-PCR or RNA-Seq data using fold change values for filtering.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.