OncoScan™ CNV Reagent Kit for Research - FAQs
OncoScan™ CNV Reagent Kit for Research - FAQs
View additional product information for OncoScan™ CNV Reagent Kit for Research - FAQs (902692TS)
17 product FAQs found
Is there an equivalent metric for the "Offset Flag" in the QC and Sample Info tab in Chromosome Analysis Suite (ChAS) for OncoScan?
There is no available equivalent to the "Offset Flag" in the QC and Sample Info tab in ChAS. The "Offset Flag" is a QC metric that is found in the Analysis Dashboard for OncoScan products. This QC metric lets us know if the TuScan algorithm detected that the usual normalization method was off, and therefore required readjustment.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What data files are produced during the assay and analysis process?
These data files are produced that are key to the process:
ARR file - This file includes sample information.
AUDIT file - This file is a log of the sample history.
DAT file - This file is the raw data from the scanner.
CEL file - This file is the gridded and processed data.
xxCHP file - This file is the output of ChAS 3.1 and contains all of the analysis data.
CHPCAR file - This file stores user-annotated calls, interpretations, and modifications made to CHP file segment data (ChAS v2.0 and higher).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the difference between OncoScanCancerGeneOnly.r1 and OncoScanGeneBoundaries.r1?
OncoScanGeneBoundaries.r1 lists the approximately 900 OncoScan cancer genes and includes greater than 10 kb on each side of the gene. OncoScanCancerGeneOnly.r1 lists the same approximately 900 OncoScan cancer genes using the start and stop positions of the gene (no additional 10 kb on each side of the gene).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What does "low diploid flag" mean?
The algorithm identifies normal diploid markers in the cancer samples. This is particularly important in highly aberrant samples. The normal diploid markers are used to calibrate the signals so that a log2 ratio of 0 (e.g., copy number 2) is achieved. In about 2% of samples, the algorithm cannot identify a sufficient number of “normal diploid” markers, and no normal diploid calibration occurs. This event triggers “low diploid flag = YES.” In this case, the user needs to carefully examine the log2 ratios and verify that re-centering is necessary.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is "celpaircheckstatus," and what does it do?
Celpaircheckstatus checks if the results from the AT channel are consistent with the results from the GC channel and represent the results of the respective channel. This metric will be “out of bounds,” for example, when the AT and GC channels were mispaired between samples or due to sample contamination. We recommend troubleshooting as described in the OncoScan Console User Manual.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
How are the somatic mutations calls made?
The ChAS software includes a table with the following calls made for each mutation: “high confidence,”, “lower confidence,” and “undetected.” The thresholds for the “high-confidence” calls were established based on separations between the normal reference and mutant calls that resulted in 95% sensitivity and 99% specificity (for lower confidence) and 95% sensitivity and 99.9% specificity (for high confidence) in our spike in experiments. The software provides for visualization of mutant probes versus reference probes representing the wild type to enable users to visually assess this separation for their sample. There is no overlay with copy number data (other than through table summaries).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What does "ploidy status" mean?
Ploidy refers to the baseline copy number observed in the tumor portion of the sample. Ploidy cannot always be estimated. For example, a sample with four copies of each chromosome and no other aberrations looks the same as a sample with two copies of each chromosome.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Under what conditions can TuScan estimate the percentage of aberrant cell fraction?
This is possible when:
The tumor is nearly homogeneous (i.e., it has a major dominant clone), and the percentage of aberrant cell fraction is 40% or higher.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What does the percentage of aberrant cells mean?
This represents the percentage of aberrant cells in a sample:
When reported as “N/A,” it means that the percentage of aberrant cells could not be estimated.
When reported as a percentage (e.g., 60%), it means that 60% of the cells were aberrant.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What copy number calls do I get from TuScan?
Log2 and B-allele frequency.
Linear copy number calls when you mouse over aberrations. In some cases, linear calls are calls in aberrant cell fraction only. In the software, when the percentage of aberrant cells has a percentage value, the linear call for aberrations in that sample is the call in the aberrant cells only. Values will be integers. When the percentage of aberrant cells is “N/A,” the linear call is an average call across the aberrant and non-aberrant cells, and values will be non-integers. You can estimate the copy number in aberrant cells if you have an independent tumor burden estimate.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What algorithm(s) can be used to make copy number calls?
We recommend the TuScan algorithm. This algorithm has fixed settings based on 19 probes/call to ensure high sensitivity and specificity at the stated resolution claims. Nexus Express Software for OncoScan FFPE Assay Kit has an additional algorithm for copy number calls from OncoScan FFPE Assay Kit: SNPFASST2 algorithm, developed and supported by BioDiscovery. This allows the user to adjust settings and make custom calls (e.g., with fewer probes). Use cases are:
When breakpoints are clear with fewer probes
When probe re-centering is needed
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Do you have exon coverage in the high-resolution genes?
Yes, if using the Affymetrix defined workflow and depending on the size of the exon (our algorithm requires 20 contiguous markers to make a call). If using an “open” workflow, the user can define the number of probes they need to claim exon resolution.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the HapMap concordance of genotyping calls?
The HapMap concordance of genotyping calls is greater than 99%.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the dynamic range of OncoScan CNV & Oncoscan CNV Plus Assay Kit?
OncoScan CNV & Oncoscan CNV Plus Assay Kit can detect copy number changes up to 50 copies. As the number of copies increases, the user should bin the calls. For example, copy numbers of 20-50 should be considered as one bin. The recommended bins are: 0, 1, 2, 3, 4, 5-6, 7-10, 11-20, 20-50.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For a specific aberration, what is the minimal resolution of the TuScan algorithm in calling a segment in terms of percentage of cells containing the aberration?
OncoScan CNV & Oncoscan CNV Plus Assay Kit can detect mosaic aberrations found in 25% of the aberrant cells. TuScan operates in two modes:
When the percentage of aberrant cell fraction is ≥40% in a homogeneous tumor, TuScan makes calls that reflect the integer copy number in the aberrant cells.
In all other cases, TuScan makes calls down to 25% aberrant cell fractions. These are “fractional” copy number calls (such as 2.3) reflecting the average CN across the aberrant and non-aberrant cell fraction.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the genome-wide resolution of loss of heterozygosity (LOH)?
The genome-wide resolution of LOH genome wide is less than or equal to 10 MB. If the aberrant cell fraction is high, the resolution increases to 3 to 5 MB.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is the resolution within cancer genes and genome-wide outside of the cancer genes?
OncoScan CNV & Oncoscan CNV Plus Assay Kit have a 50 kb-100 kb resolution in approximately 900 cancer genes.
Outside of the cancer genes:
88% of the genome has 300 kb resolution
97% of the genome has at least 380 kb resolution
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.