Product Image
Applied Biosystems™
Axiom™ Microbiome Reagent Kit for four 24-array plates
The Axiom Microbiome Reagent Kit is used with the Axiom Microbiome 24-Array or 96-Array Plates. The Axiom Microbiome Array Plates enables the detection of all known microorganisms in a sample with species- and strain-level identification in a single, scalable assay.
Have Questions?
Catalog NumberQuantity
9029101 kit
Catalog number 902910
Price (USD)
-
Quantity:
1 kit
The Axiom Microbiome Reagent Kit is used with the Axiom Microbiome 24-Array or 96-Array Plates. The Axiom Microbiome Array Plates enables the detection of all known microorganisms in a sample with species- and strain-level identification in a single, scalable assay.
The Reagent Kit contains all reagents (except isopropanol) to process four 24-array plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 kit
Reagent Typemicrobial processing reagents
TypeMicrobiome Reagent Kit
Unit SizeEach
Frequently asked questions (FAQs)
Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?
The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.
What is the starting amount of DNA required for Axiom arrays?
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.