Clariom™ S Pico Assay HT, mouse, 30 reactions - FAQs
Clariom™ S Pico Assay HT, mouse, 30 reactions - FAQs
View additional product information for Clariom™ S Pico Assay HT, mouse - FAQs (902965, 902966)
1 product FAQs found