Clariom™ S Assay HT, rat
Applied Biosystems™

Clariom™ S Assay HT, rat

Obtain a gene-level view of the rat transcriptome with the Clariom S Assays HT for rat. The Clariom S Assays,Read more
Have Questions?
Catalog NumberNumber of Arrays
90297324 arrays
Catalog number 902973
Price (USD)
5,010.00
Each
Number of Arrays:
24 arrays
Price (USD)
5,010.00
Each
Obtain a gene-level view of the rat transcriptome with the Clariom S Assays HT for rat. The Clariom S Assays, rat serve as a next generation transcriptome-wide gene-level expression profiling tool, which allow for the fastest, simplest, and most scalable path to generating actionable results. Based on industry-leading microarray technology, the novel rat HT Clariom S Assay design provides extensive coverage of all known well-annotated genes, compatibility with clinical sample types, scalable formats, and flexible data analysis software. Clariom S Assays HT are the tools of choice to find expression biomarkers with known function as quickly, easily, and cost-effectively as possible.

Find answers, move on
Although the number of known transcribed genes has expanded rapidly in recent years, knowledge of the function of each gene is still evolving. Many genes and transcripts found in databases are poorly annotated or unannotated, which can complicate and prolong data analysis and interpretation. rat HT Clariom S Assays focus on well-annotated genes, providing researchers with the ability to perform gene-level expression profiling studies and to quickly assess changes in key genes and pathways. With less time required for data analysis, Clariom S Assays HT for rat help researchers reach conclusions more rapidly.

Simple, swift biomarker discovery
• Accurately measure gene-level expression from >20,000 well-annotated genes to get to answers quickly.
• Choose a format that suits your throughput needs, processing from 1 to 192 samples a day.
• Go from data to insight in minutes with intuitive, highly visual, free analysis software designed for the biologist.

When you have precious samples, get it right the first time
• Generate robust expression profiles from as little as 100 pg of total RNA–as few as 10 cells.
• Utilize RNA from various sample types including blood, cells, and fresh/fresh-frozen or FFPE tissues.
• Preserve sample integrity and reduce data variability with an assay that does not require a globin or rRNA removal step.
• Save time and money with fully automated sample preparation options.

Clariom S solutions are available formats for single-sample (cartridge array) processing on the GeneChip™ 3000 instrument system and high-throughput automated processing (plate array) on the GeneTitan™ HT Microarray System, offering the flexibility to accommodate both small and large cohort studies. The complete solution comes with reagents and fast, simple Transcriptome Analysis Console (TAC) software to analyze and visualize global expression patterns of genes, pathways, and network interactions in minutes.

Get the truest level of gene-level expression
To generate robust gene-level expression, rat HT Clariom S Assays detect only the exons present in all known transcript isoforms expressed from a single gene locus-constitutive exons. This differs from other gene-level array technologies and shallow RNA-Seq, which provide either a biased view of gene expression or data that are complicated by variation in expression of transcript variants. By detecting only constitutive exons throughout the length of each known gene, rat HT Clariom S Assays generate the most accurate and truest measurement of gene-level expression available today.

Keeping biomarker identification across the transcriptome simple and swift, Clariom S Assays HT for rat provide you with the coverage you require, the reproducibility you need, and the insights you want to act on your discoveries.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ArrayTranscriptome Profiling
Array Format24-array Plate
For Use With (Application)Microarray Analysis
No. of Reactions30
Number of Arrays24 arrays
Product LineApplied Biosystems
Product TypeS Assay HT
Quantity30 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
SpeciesRat
Sample TypeRNA
Unit SizeEach
Contents & Storage
• Poly-A RNA Control Kit, store at -20°C
• Hybridization Control Kit, store at -20°C
• WT Terminal Labeling Kit, store at -20°C
• WT Amplification Kit, Module 1, store at -20°C
• WT Amplification Kit, Module 2, store at 4°C
• Clariom S 24-Array Plate, rat, store at 4°C

Frequently asked questions (FAQs)

What reagent kit should I use with my array?

Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I install Expression Console Software and TAC Software library files for human, mouse, or rat transcriptome assays?

Analysis library files and annotation files can be downloaded from within the TAC Software. Please refer to the respective user manuals for detailed instructions.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What software is available for analyzing the results from my human, mouse, or rat transcriptome assays?

Probe cell intensity data (CEL) from human, mouse, or rat transcriptome assays are analyzed in Transcriptome Analysis Console(TAC) Software. The application uses a version of RMA analysis to create CHP files. Transcriptome Analysis  Console Software can be downloaded at no charge from the website. Further statistical analysis may be performed in Affymetrix' Transcriptome Analysis Console (TAC) Software to obtain a list of differentially expressed genes and alternative splicing events. TAC Software also provides the visualization of genes, exons, junctions, transcript isoforms, pathways, and mRNA-miRNA regulatory networks.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I install AGCC library files for the human, mouse, or rat transcriptome arrays?

To install library files for the human, mouse, or rat transcriptome arrays for AGCC, simply run the executable library file installer.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What format are the human, mouse, or rat transcriptome arrays?

Human, mouse, or rat transcriptome arrays are 49-format.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.