Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, single door, holds 240 boxes, 120V/60Hz, 16A - FAQs
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, single door, holds 240 boxes, 120V/60Hz, 16A - FAQs
View additional product information for Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (903)