CD8 APC-eFluor™ 780 SK1, CE-IVD, eBioscience™ - FAQs
CD8 APC-eFluor™ 780 SK1, CE-IVD, eBioscience™ - FAQs
View additional product information for CD8 APC-eFluor™ 780 SK1, CE-IVD, eBioscience™ - FAQs (9047-0087-120)
1 product FAQs found
Can the eFluor Organic Dyes be frozen?
As with other organic fluorochromes, we do not recommend the eFluor Organic Dyes be frozen.