Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, Capacity: 17.3 cu. ft. (489.9L) capacity, single door, holds 320 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, Capacity: 17.3 cu. ft. (489.9L) capacity, single door, holds 320 boxes, 230V/50Hz, 12A, painted galvaneel interior - FAQs
View additional product information for Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (905)