KaryoStat™ Assay
Applied Biosystems™

KaryoStat™ Assay

The KaryoStat Assay is an alternative to g-banding karyotyping offering accurate genotyping (sample ID) plus whole-genome coverage for accurate detectionRead more
Have Questions?
Catalog NumberQuantity
90540324 assays
Catalog number 905403
Price (USD)
-
Quantity:
24 assays
The KaryoStat Assay is an alternative to g-banding karyotyping offering accurate genotyping (sample ID) plus whole-genome coverage for accurate detection of chromosomal abnormalities with results in 2.5 days. The assay enables the detection of aneuploidies, submicroscopic aberrations, and mosaic events for research use. Because the assay provides genotype information, it can help in the identification of triploidies and copy neutral events such as absence of heterozygosity. Our proprietary manufacturing technology produces arrays that are highly reproducible between each batch with no risk of probe dropout inherent in bead manufacturing techniques. The assay includes arrays, reagents, and easy-to-use data analysis software for a cost-effective and streamlined analysis of pluripotent stem cell lines.

Features include:
• Cost effective and accurate detection of chromosomal abnormalities
• Karyotyping and genotyping (sample ID) with one assay
• Simple analysis; no cytogenetics expertise necessary
• Assay includes arrays, reagents, and free downloadable software
• 2.5 days to results

 KaryoStat AssayKaryoStat HD Assay
Gains>2 Mb>25–50 kb
Losses>1 Mb>25–50 kb
Absence of heterozygosity>5 Mb>1 Mb
SNPYesYes
CoverageLowHighest
InstrumentGeneChip Scanner 3000 onlyGeneChip Scanner 3000 only
Analysis softwareChromosomal Analysis SuiteChromosomal Analysis Suite
Specifications
IncludesArrays, Reagents, Data Analysis Software
Product LineKaryoStat
Quantity24 assays
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
TypeAssay
FormatGenechip Probe Array
Number of Arrays24 arrays
SpeciesHuman
Unit SizeEach
Contents & Storage
Store at 2–8°C:
• Arrays
• Module 2: Pre-Lab TE Buffer and Water
• Module 4: Main Lab Stain, Holding Buffer, Beads, and Water

Store at -5 to -30°C:
• Module 1: Pre-Lab Restriction, Ligation, and Amplification
• Module 3: Main Lab Fragmentation, Labeling, and Hybridization

Store at room temperature:
• Module 5: Main Lab Elution Buffer
• Purification Wash Buffer
• Wash Buffer A
• Wash Buffer B