GeneChip™ PrimeView™ Global Gene Expression Profile Assay, 30 reactions, 30 arrays - FAQs
GeneChip™ PrimeView™ Global Gene Expression Profile Assay, 30 reactions, 30 arrays - FAQs
View additional product information for GeneChip™ PrimeView™ Global Gene Expression Profile Assay - FAQs (905407, 905406)
2 product FAQs found
Which kit do I need to purchase to run the PluriTest Assay?
Customers should purchase the GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905406 or 905407). 
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What additional reagents do I need to run the GeneChip PrimeView Global Gene Expression Profile Assay, Cat. No. 905406 or 905407?
The GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905406 or 905407) does not include the hybridization, wash, and stain reagents. Hence, the GeneChip Hybridization, Wash, and Stain Kit (Cat. No. 900720) will have to be purchased separately. Alternatively, the GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905400) can be used; it is a complete kit that includes the GeneChip Hybridization, Wash, and Stain kit. 
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.