Axiom™ Asia Precision Medicine Research Array Kit, 96-format - FAQs
Axiom™ Asia Precision Medicine Research Array Kit, 96-format - FAQs
View additional product information for Axiom™ Asia Precision Medicine Research Array Kit, 96-format - FAQs (905423)
3 product FAQs found
What is the starting amount of DNA required for Axiom arrays?
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the pre-amplification clean room requirements for Axiom assays?
When manipulating genomic DNA, precautions need to be taken to avoid contamination with foreign DNA amplified in other reactions and procedures.
- We recommend that genomic DNA manipulations be performed in a dedicated pre-amplification room or area separate from the main laboratory. This pre-amplification area should have a dedicated set of pipettes and plasticware.
- If a dedicated area is not available, we recommend using a dedicated bench or a dedicated biosafety hood with dedicated pipettes.
- If a dedicated bench or biosafety hood is not available, a set of dedicated pipettes is recommended at the least.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.