Product Image
Applied Biosystems™

UK Biobank Axiom™ Array Kit

The UK Biobank Axiom Array was designed by and for thought leaders in the human genetics community for high-throughput, high-valueWeitere Informationen
Have Questions?
KatalognummerMenge
905430jeweils 1
Katalognummer 905430
Preis (EUR)
-
Menge:
jeweils 1
The UK Biobank Axiom Array was designed by and for thought leaders in the human genetics community for high-throughput, high-value genotyping of large sample cohorts to explore the genetics of complex diseases and translational research with a single comprehensive low-cost solution. The 96-format array plate allows for genome-wide genotyping of large sample collections such as those screened at biobanks, genome centers, and core labs. This array kit includes the 1x96-format array plate, as well as an Axiom 2.0 Reagent Kit and Axiom GeneTitan Consumables Kit.

GWAS
• Intelligent marker selection enables imputation of millions of additional SNPs
• The genomic content module of each array is optimized for specific populations and is customizable to any 1000 Genomes population study by adding additional markers of your choosing

Pharmacogenomic markers
• Markers were selected to represent phases of absorption, distribution, metabolism, and excretion (ADME)

Inflammation and HLA
• Contains markers with evidence for association with autoimmune and inflammation
• Covers variants in genes in the HLA and KIR regions known to be important in immune response

Exome
• Rare, non-synonymous coding SNPs and indels in protein coding regions of the genome
• A majority of variants are rare with minor allele frequency (MAF) <1%
• Newly discovered loss-of-function content
• SNPs and indels identified from a sequencing initiative of 26,000 individuals
• Known disease-causing mutations

eQTLs
• Markers that have known associations to RNA expression traits
• Captures unique eQTLs contained in the NCBI Genotype-Tissue Expression (GTEx) eQTL database, GEUVIDAS project, and several other discovery projects

Human disease
• Alzheimer-s disease, including coverage of ApoE
• Blood phenotypes
• Cancer common variants
• Cardiometabolic

Choose your content
• Order the pre-designed array and customize it with markers of your choice
• Custom markers may be de novo or selected from the Axiom Genomic Database of genotype-tested markers

Please note: Reagent kits do not include plastic consumables required to run the assay on the Beckman Biomek™ FXP Target Prep Express System. Please contact us for a list of required plastic consumables.
Specifications
Zur Verwendung mit (Anwendung)Microarray-Analyse
ProduktlinieAxiom
Mengejeweils 1
VersandbedingungApproved for shipment at Room Temperature or on Wet or Dry Ice
TypArray Kit
ArrayGenotyping
Format96-array Plate
Anzahl Arrays96 Arrays
SpeziesHuman
Unit SizeEach
Inhalt und Lagerung
• 1x96-format array plate. Store at 2–8°C.
• 1 Axiom 2.0 Reagent Kit (includes reagents and controls for 1x96-format array plate). Storage can vary for each reagent; refer to product insert.
• 1 Axiom GeneTitan Consumables Kit (includes GeneTitan instrument consumables to process 1x96 array plate. Store at room temperature.

Häufig gestellte Fragen (FAQ)

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.