Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Thermo Scientific™
Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFilteredFit for PurposeUnit Size
906-26250 μLNon-sterileNon-filteredCertified QualityCase of 4800
906-02150 μLSterileFilteredSterile and PCR ReadyCase of 4800
906-021PK
also known as 906-021
50 μLSterileFilteredSterile and PCR ReadyPack of 960
906-26150 μLSterileNon-filteredSterileCase of 4800
906-261PK
also known as 906-261
50 μLSterileNon-filteredSterilePack of 960
907-02180 μLSterileFilteredSterile and PCR ReadyCase of 4800
907-021PK
also known as 907-021
80 μLSterileFilteredSterile and PCR ReadyPack of 960
907-261100 μLSterileNon-filteredSterileCase of 4800
907-261PK
also known as 907-261
100 μLSterileNon-filteredSterilePack of 960
907-262100 μLNon-sterileNon-filteredCertified QualityCase of 4800
907-262PK
also known as 907-262
100 μLNon-sterileNon-filteredCertified QualityPack of 960
908-021150 μLSterileFilteredSterile and PCR ReadyCase of 4800
908-021PK
also known as 908-021
150 μLSterileFilteredSterile and PCR ReadyPack of 960
908-261200 μLSterileNon-filteredSterileCase of 4800
908-261PK
also known as 908-261
200 μLSterileNon-filteredSterilePack of 960
908-262200 μLNon-sterileNon-filteredCertified QualityCase of 4800
908-262PK
also known as 908-262
200 μLNon-sterileNon-filteredCertified QualityPack of 960
Catalog number 906-262
Price (USD)
571.50
Online exclusive
635.00
Save 63.50
Case of 4800
Volume (Metric):
50 μL
Sterility:
Non-sterile
Filtered:
Non-filtered
Fit for Purpose:
Certified Quality
Unit Size:
Case of 4800
Price (USD)
571.50
Online exclusive
635.00
Save 63.50
Case of 4800
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius
FormatRack
Product LineTecan
SterilityNon-sterile
Tip StyleLow Retention, Extended Length
Volume (Metric)50 μL
Unit SizeCase of 4800