Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, 15mL Ampule - FAQs
Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezer, 15mL Ampule - FAQs
View additional product information for Forma™ 900 Series -86°C Upright Ultra-Low Temperature Freezers - FAQs (910)