Orion™ pH Buffer Cubitainers™
Thermo Scientific™

Orion™ pH Buffer Cubitainers™

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH
9104CB4.01
9107CB7
9110CB10.01
Catalog number 9104CB
Price (USD)
188.10
Online exclusive
209.00
Save 20.90
Each
pH:
4.01
Price (USD)
188.10
Online exclusive
209.00
Save 20.90
Each

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
  • Manufactured under ISO 9000 quality standards
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Boiling Point100°C
Certificate of AnalysisYes
Certifications/ComplianceISO 9001
Chemical Name or MaterialpH Buffer, 5 gallon cubitainers
ColorLight Red
DescriptionpH 4.01 Buffer, 5 gallon
FormulationWater, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
PackagingCubitainer
Solubility InformationSoluble
Traceability to NISTYes
For Use With (Application)Accurate, reproducible pH calibrations
For Use With (Equipment)pH Meter and Electrode
GradeCertified
Physical FormLiquid
Quantity19 L
Solution TypepH Buffer
pH4.01
Unit SizeEach