Orion™ Single Junction Epoxy Bodied pH/ATC Triode™
Thermo Scientific™

Orion™ Single Junction Epoxy Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesLength (Metric) Cable
9107WMMDBNC (Waterproof)3 m
9107BNBNC1 m
9107BNMDBNC (Waterproof)1.5 m
Catalog number 9107WMMD
Price (USD)
360.00
Online exclusive
400.00
Save 40.00
Each
Connector Types:
BNC (Waterproof)
Length (Metric) Cable:
3 m
Price (USD)
360.00
Online exclusive
400.00
Save 40.00
Each

Thermo Scientific™ Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes are low-maintenance, gel-filled and come with a built-in ATC. They are engineered with an epoxy body for ruggedness and best suited for general purpose samples or for educational institutes where minimal maintenance is required.

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
Specifications
DescriptionpH/ATC Triode
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
For Use With (Application)The Thermo Scientific Orion 9107BN Triode 3-in-1 pH/automatic temperature compensation probe has a low maintenance gel filled electrode with built-in ATC.For field or plant use.
For Use With (Equipment)Dual Star, Orion STAR A111, Orion STAR A121, Orion STAR A211, Orion STAR A221, Orion STAR A321, Versa Star
JunctionWick
Length (English)4.7 in.
Length (Metric)120 mm
Length (Metric) Cable3 m
MaterialEpoxy
Reference TypeAg/AgCl
Temperature (English) Operating32°F to 176°F
Connector TypesBNC (Waterproof)
Temperature (Metric) Operating0°C to 80°C
TypepH/ATC Probe, BNC Waterproof Connector
pH Range0 to 14
Unit SizeEach