Orion™ Double Junction Semi-Micro Stem Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ Double Junction Semi-Micro Stem Glass Bodied Combination pH Electrode

Top-performance, semi-micro pH electrodes to measure samples as small as 0.2 mL.
Have Questions?
Catalog NumberConnector Types
9110DJWPBNC (Waterproof)
Catalog number 9110DJWP
Price (USD)
318.60
Online exclusive
354.00
Save 35.40
Each
Connector Types:
BNC (Waterproof)
Price (USD)
318.60
Online exclusive
354.00
Save 35.40
Each
Thermo Scientific™ Orion™ Double Junction Semi-Micro Electrodes provide reliable and accurate measurements of routine samples. They are available in chemically resistant glass bodies and best suited for measuring general samples as small as 0.2mL, in test tubes and small containers.
 

Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples.

Solutions required

  • Electrode fill solution
  • 910008 3M KCl filling solution, 5 x 60 mL bottles

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
  • electrode storage solution, 475 mL, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 910008 3M KCl filling solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase
Specifications
DescriptionDouble Junction pH Electrode
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter
Internal ReferenceAg/AgCl Double Junction
Precision0.02
Temperature (English) Operating32°F to 140°F
Connector TypesBNC (Waterproof)
JunctionCeramic
MaterialGlass Body
Temperature (Metric) Operating0°C to 60°C
TypeDouble Junction pH Electrode
pH Range0 to 14
Unit SizeEach