Orion™ Single Junction Combination pH Electrodes
Thermo Scientific™

Orion™ Single Junction Combination pH Electrodes

Conduct routine, small volume or flat surface pH measurement with Thermo Scientific™ Orion™ economy series pH electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesTypeTip Type
912600BNCCombination pH ElectrodeFlask
9106BNWPBNC (Waterproof)Combination pH ElectrodeStandard
911600BNCCombination pH ElectrodeSemi-micro
913600BNCCombination pH ElectrodeFlat Surface
Catalog number 912600
Price (USD)
375.30
Online exclusive
417.00
Save 41.70
Each
Connector Types:
BNC
Type:
Combination pH Electrode
Tip Type:
Flask
Price (USD)
375.30
Online exclusive
417.00
Save 41.70
Each
Thermo Scientific™ Orion™ Low Maintenance pH Electrodes provide reliable and accurate measurements of routine samples. 9106BNWP is best suited for field or plant use with general purpose samples, and educational institutes where minimal maintenance is required. 911600 is ideal for samples as small as 0.2mL. 912600 is suited for use with long neck flask. 913600 has a flat sensing bulb good for measurement of flat, soft, moist, semi-solid or small volume samples.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
Specifications
DescriptionPermanently gelled Ag/AgCl electrolyte
Diameter (English) Body0.25 in.
Diameter (English) Cap0.18 in.
Diameter (Metric) Body6 mm
Diameter (Metric) Cap10 mm
For Use With (Equipment)Standard pH Meter
Length (English)12.01 in.
Length (Metric)30.5 cm
Length (Metric) Cable1 m
Reference ElectrolyteAg/AgCl
RefillableNon-refillable
Temperature (English) Operating32°F to 176°F
Tip TypeFlask
Connector TypesBNC
JunctionWick
MaterialEpoxy
Temperature (Metric) Operating0°C to 80°C
TypeCombination pH Electrode
pH Range0 to 12
Unit SizeEach