Evolution™ Pro UV-Vis Spectrophotometers
Thermo Scientific™
Evolution™ Pro UV-Vis Spectrophotometers
Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.
Have Questions?
變更視圖buttonViewtableView
產品號碼Type
912A1113Evolution Pro INTL USP/EP Standards Bundle
840-340200UV-Vis Spectrophotometer
840-340100UV-Vis Spec w/ Mercury Lamp Bundle
912A1110Evolution Pro US Pharma Value Bundle
912A1112Evolution Pro INTL Pharma Value Bundle
840-372600Evolution Pro + Mercury Lamp US Pharma Value Bundle
840-372800Evolution Pro + Mercury Lamp INTL Pharma Value Bundle
912A1111Evolution Pro US USP/EP Standards Bundle
840-372700Evolution Pro + Mercury Lamp US USP/EP Standards Bundle
840-372900Evolution Pro + Lamp INTL USP/EP Standards Bundle
產品號碼 912A1113
價格 (TWD)
-
Type:
Evolution Pro INTL USP/EP Standards Bundle
For routine, complex research and advanced applications, including current compliance, the Thermo Scientific Evolution Pro UV-Vis Spectrophotometer series brings together reliable, versatile hardware, a broad range of accessories, with easy-to-use, easy-to-learn Thermo Scientific Insight Pro Software.
  • Easily go from sample to final report - with intuitive INSIGHT Pro Software guiding you every step of the way.
  • Obtain results you need for quantitative analysis, scanning and kinetics applications with comprehensive software tools for data collection, processing, and reporting.
  • Customize even complex methods with ease using workflow-oriented application modules.
  • Enforce data security with optional INSIGHT Pro Security Software tools to assist with achieving current 21 CFR Part 11 compliance in your pharmaceutical lab.
Enhance productivity with sampling flexibility:
  • Speed up your analysis with Thermo Scientific™ Smart Accessories with snap-in design for convenience and consistency.
  • Eliminate manual setup with auto-recognition and software integration of sippers, cells changers, and auto-sampler accessories.
  • Take advantage of the Evolution Pro UV-Vis Spectrophotometer’s wide beam separation and large sample compartment, providing flexibility for unique accessories and advanced applications.
Regulatory/Quality Control Environments:
To use the Evolution Pro UV-Vis Spectrophotometer in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.
Evolution Pro Pharma Value Bundles include:
Evolution Pro Spectrometer
INSIGHT Pro with Security Software 
Non-USP and EP Standards Set
Evolution Pro USP/EP Standards Pharma Bundles include:
Evolution Pro Spectrometer
INSIGHT Pro with Security Software
USP and EP UV Standards Set
USP and EP Visible Standards Set
Accessory Ordering Information:
All accessories are sold separately. Other accessories include cell changers, cuvette and test holders, calibration carousels, sippers, fiber optic probes, small volume sampling, and accessories for reflectance and rapid mixing.
規格
IncludesEvolution Pro UV-Vis Spectrometer, INSIGHT Pro with Security Software, Validator for Insight Pro A4 Paper Version, USP and EP UV Standards Set, USP and EP Visible Standards Set
Product LineEvolution Pro
Spectral Bandwidth0.5, 1.0, 1.5, 2.0, 4.0 nm Selectable
TypeEvolution Pro INTL USP/EP Standards Bundle
Unit SizeEach
常見問答集 (常見問題)
Who should perform service and calibration on Evolution instruments?
Alignment, internal calibration, and service procedures should be performed by authorized Thermo Fisher Scientific service personnel.
How is performance verified with the Evolution instruments?
Performance verification is performed using built-in routines and certified standards according to the user guide and laboratory qualification procedures. Evolution instruments also comply with USP <857> and Ph. Eur. 2.2.25, and include performance verification testing for ensuring performance within those guidelines.
Are there models optimized for life science applications on the Evolution?
Yes. Insight Pro software includes preconfigured methods for DNA, RNA, and protein quantification, including popular assays like Protein Bradford.
Are kinetics and enzyme rate studies supported with Evolution instruments?
Yes. Evolution instruments support advanced kinetics, including multi-wavelength kinetics and time-based studies for enzymatic and reaction monitoring applications.
Can the Evolution family support automation?
Yes. Available accessories include:
  • Automatic multi-cell changers
  • Sipper systems
  • Autosamplers